Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119105597-119105858 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLEC10
Entrez Gene ID
10584
Gene Name
collectin subfamily member 10
Gene Aliases
CL-34, CLL1
UniGene
Hs.176615
Gene Chromosome Location
Chr.8: 118952167-119108455 on Build GRCh38
Transcripts
NM_001324095.1 NM_006438.4 XM_005250756.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1454482 c.486C>T p.I162I Chr.8: 119105843-119105843 on Build GRCh38
1095430 c.477C>A p.F159L Chr.8: 119105834-119105834 on Build GRCh38
5601299 c.460G>A p.E154K Chr.8: 119105817-119105817 on Build GRCh38
4548343 c.444G>A p.V148V Chr.8: 119105801-119105801 on Build GRCh38
748493 c.477C>T p.F159F Chr.8: 119105834-119105834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375600625 Chr.8: 119105755-119105755 on Build GRCh38
rs753449295 Chr.8: 119105772-119105772 on Build GRCh38
rs754480680 Chr.8: 119105777-119105777 on Build GRCh38
rs187243796 Chr.8: 119105648-119105648 on Build GRCh38
rs927420654 Chr.8: 119105765-119105765 on Build GRCh38
rs1021011665 Chr.8: 119105795-119105795 on Build GRCh38
rs1049308134 Chr.8: 119105718-119105718 on Build GRCh38
rs757232356 Chr.8: 119105819-119105819 on Build GRCh38
rs946045725 Chr.8: 119105825-119105825 on Build GRCh38
rs905486038 Chr.8: 119105729-119105729 on Build GRCh38
rs777502820 Chr.8: 119105796-119105796 on Build GRCh38
rs758132833 Chr.8: 119105794-119105794 on Build GRCh38
rs752119091 Chr.8: 119105769-119105769 on Build GRCh38
rs202199030 Chr.8: 119105770-119105770 on Build GRCh38
rs555457969 Chr.8: 119105716-119105716 on Build GRCh38
rs368636526 Chr.8: 119105779-119105779 on Build GRCh38
rs781073829 Chr.8: 119105823-119105823 on Build GRCh38
rs11987106 Chr.8: 119105782-119105782 on Build GRCh38
rs746970099 Chr.8: 119105809-119105809 on Build GRCh38
rs149406487 Chr.8: 119105643-119105643 on Build GRCh38
rs574300596 Chr.8: 119105724-119105724 on Build GRCh38
rs779456796 Chr.8: 119105656-119105656 on Build GRCh38
rs78345659 Chr.8: 119105693-119105693 on Build GRCh38

View Full Product Details