Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158292011-158292270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897690 c.378G>T p.K126N Chr.1: 158292133-158292133 on Build GRCh38
1688837 c.331C>T p.P111S Chr.1: 158292086-158292086 on Build GRCh38
897691 c.393C>A p.F131L Chr.1: 158292148-158292148 on Build GRCh38
5823912 c.508T>A p.Y170N Chr.1: 158292263-158292263 on Build GRCh38
1688838 c.511G>A p.E171K Chr.1: 158292266-158292266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764463540 Chr.1: 158292097-158292097 on Build GRCh38
rs765694898 Chr.1: 158292145-158292147 on Build GRCh38
rs756038893 Chr.1: 158292220-158292220 on Build GRCh38
rs1045058762 Chr.1: 158292239-158292239 on Build GRCh38
rs761179568 Chr.1: 158292061-158292061 on Build GRCh38
rs757907860 Chr.1: 158292088-158292088 on Build GRCh38
rs776508449 Chr.1: 158292039-158292039 on Build GRCh38
rs750112457 Chr.1: 158292187-158292187 on Build GRCh38
rs779059996 Chr.1: 158292114-158292114 on Build GRCh38
rs750070786 Chr.1: 158292087-158292087 on Build GRCh38
rs777071590 Chr.1: 158292137-158292138 on Build GRCh38
rs749540015 Chr.1: 158292040-158292040 on Build GRCh38
rs201448758 Chr.1: 158292108-158292108 on Build GRCh38
rs368146889 Chr.1: 158292053-158292053 on Build GRCh38
rs765826564 Chr.1: 158292195-158292195 on Build GRCh38
rs867887479 Chr.1: 158292092-158292092 on Build GRCh38
rs772050553 Chr.1: 158292127-158292127 on Build GRCh38
rs111577378 Chr.1: 158292109-158292109 on Build GRCh38
rs777045453 Chr.1: 158292167-158292167 on Build GRCh38
rs1005150918 Chr.1: 158292129-158292129 on Build GRCh38
rs751036560 Chr.1: 158292198-158292198 on Build GRCh38
rs370791324 Chr.1: 158292057-158292057 on Build GRCh38
rs775584477 Chr.1: 158292132-158292132 on Build GRCh38
rs1012327169 Chr.1: 158292101-158292101 on Build GRCh38
rs115959450 Chr.1: 158292098-158292098 on Build GRCh38
rs754910860 Chr.1: 158292107-158292107 on Build GRCh38
rs769112361 Chr.1: 158292166-158292166 on Build GRCh38
rs147598183 Chr.1: 158292072-158292072 on Build GRCh38
rs762204489 Chr.1: 158292140-158292141 on Build GRCh38
rs868151001 Chr.1: 158292086-158292086 on Build GRCh38
rs771234067 Chr.1: 158292051-158292051 on Build GRCh38
rs771222792 Chr.1: 158292206-158292206 on Build GRCh38
rs956911228 Chr.1: 158292048-158292048 on Build GRCh38
rs148953286 Chr.1: 158292193-158292193 on Build GRCh38
rs375298399 Chr.1: 158292200-158292200 on Build GRCh38
rs778750254 Chr.1: 158292140-158292140 on Build GRCh38
rs777559090 Chr.1: 158292233-158292233 on Build GRCh38
rs560975462 Chr.1: 158292096-158292096 on Build GRCh38
rs939843817 Chr.1: 158292148-158292148 on Build GRCh38
rs746097005 Chr.1: 158292119-158292119 on Build GRCh38
rs765575548 Chr.1: 158292183-158292183 on Build GRCh38
rs575932648 Chr.1: 158292066-158292066 on Build GRCh38
rs781327520 Chr.1: 158292203-158292203 on Build GRCh38
rs759582146 Chr.1: 158292055-158292055 on Build GRCh38
rs764913992 Chr.1: 158292074-158292074 on Build GRCh38
rs761108555 Chr.1: 158292133-158292133 on Build GRCh38
rs1053882181 Chr.1: 158292070-158292070 on Build GRCh38
rs754153474 Chr.1: 158292069-158292069 on Build GRCh38

View Full Product Details