Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104898275-104898546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP12
Entrez Gene ID
100506742
Gene Name
caspase 12 (gene/pseudogene)
Gene Aliases
CASP-12, CASP12P1
UniGene
Hs.476989
Gene Chromosome Location
Chr.11: 104885718-104898670 on Build GRCh38
Transcripts
NM_001191016.2 NR_034061.2 NR_034063.2 NR_034064.2 NR_034065.2 NR_034066.2 NR_034067.2 NR_034068.2 NR_034070.2 NR_034071.2

SNP Detail

SNP ID Chromosome Location
rs1041656493 Chr.11: 104898443-104898443 on Build GRCh38
rs748789690 Chr.11: 104898469-104898469 on Build GRCh38
rs1011409920 Chr.11: 104898431-104898431 on Build GRCh38
rs900381749 Chr.11: 104898323-104898323 on Build GRCh38
rs753852892 Chr.11: 104898411-104898411 on Build GRCh38
rs534751749 Chr.11: 104898338-104898338 on Build GRCh38
rs1027294325 Chr.11: 104898407-104898407 on Build GRCh38
rs938111856 Chr.11: 104898403-104898403 on Build GRCh38
rs562690583 Chr.11: 104898489-104898489 on Build GRCh38
rs1024502999 Chr.11: 104898432-104898432 on Build GRCh38
rs181033425 Chr.11: 104898305-104898305 on Build GRCh38
rs946069987 Chr.11: 104898515-104898515 on Build GRCh38
rs953125637 Chr.11: 104898484-104898484 on Build GRCh38
rs905299082 Chr.11: 104898379-104898379 on Build GRCh38
rs73629701 Chr.11: 104898447-104898447 on Build GRCh38
rs769197076 Chr.11: 104898366-104898366 on Build GRCh38
rs949398815 Chr.11: 104898319-104898319 on Build GRCh38
rs576679405 Chr.11: 104898472-104898472 on Build GRCh38
rs1030477225 Chr.11: 104898482-104898482 on Build GRCh38
rs1007728840 Chr.11: 104898498-104898498 on Build GRCh38
rs534188053 Chr.11: 104898361-104898361 on Build GRCh38
rs980276335 Chr.11: 104898462-104898462 on Build GRCh38
rs892968943 Chr.11: 104898426-104898426 on Build GRCh38
rs990381771 Chr.11: 104898488-104898488 on Build GRCh38
rs913215302 Chr.11: 104898504-104898504 on Build GRCh38
rs574783729 Chr.11: 104898325-104898325 on Build GRCh38
rs757569355 Chr.11: 104898420-104898420 on Build GRCh38

View Full Product Details