Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50671061-50671300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIP2B
Entrez Gene ID
57609
Gene Name
disco interacting protein 2 homolog B
Gene Aliases
-
UniGene
Hs.505516
Gene Chromosome Location
Chr.12: 50504985-50748667 on Build GRCh38
Transcripts
NM_173602.2 XM_005269044.1 XM_006719520.3 XM_006719521.3 XM_011538594.2 XM_011538595.2 XM_011538596.2 XM_017019699.1 XM_017019700.1 XM_017019701.1 XM_017019702.1 XM_017019703.1 XM_017019704.1 XM_017019705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4426577 c.533C>G p.S178* Chr.12: 50671291-50671291 on Build GRCh38
6644148 c.529C>G p.R177G Chr.12: 50671287-50671287 on Build GRCh38
4847646 c.445G>A p.E149K Chr.12: 50671203-50671203 on Build GRCh38
416213 c.538C>T p.Q180* Chr.12: 50671296-50671296 on Build GRCh38
4877645 c.438G>A p.S146S Chr.12: 50671196-50671196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs990188835 Chr.12: 50671148-50671148 on Build GRCh38
rs780653424 Chr.12: 50671263-50671263 on Build GRCh38
rs781297485 Chr.12: 50671167-50671167 on Build GRCh38
rs780741053 Chr.12: 50671188-50671188 on Build GRCh38
rs371530693 Chr.12: 50671155-50671155 on Build GRCh38
rs748022912 Chr.12: 50671117-50671117 on Build GRCh38
rs759527287 Chr.12: 50671229-50671229 on Build GRCh38
rs763236143 Chr.12: 50671196-50671196 on Build GRCh38
rs756155290 Chr.12: 50671255-50671255 on Build GRCh38
rs200749384 Chr.12: 50671224-50671224 on Build GRCh38
rs904963417 Chr.12: 50671165-50671165 on Build GRCh38
rs764377098 Chr.12: 50671225-50671225 on Build GRCh38
rs751783633 Chr.12: 50671235-50671235 on Build GRCh38
rs369968523 Chr.12: 50671212-50671212 on Build GRCh38
rs979197395 Chr.12: 50671243-50671243 on Build GRCh38
rs754770970 Chr.12: 50671163-50671163 on Build GRCh38
rs190447856 Chr.12: 50671116-50671116 on Build GRCh38
rs760082237 Chr.12: 50671098-50671098 on Build GRCh38
rs372754841 Chr.12: 50671234-50671234 on Build GRCh38
rs201446235 Chr.12: 50671271-50671271 on Build GRCh38
rs567723801 Chr.12: 50671253-50671253 on Build GRCh38
rs1034891653 Chr.12: 50671186-50671186 on Build GRCh38
rs763802073 Chr.12: 50671139-50671139 on Build GRCh38
rs774155722 Chr.12: 50671140-50671140 on Build GRCh38
rs767925129 Chr.12: 50671206-50671207 on Build GRCh38
rs776085981 Chr.12: 50671253-50671261 on Build GRCh38
rs373975269 Chr.12: 50671164-50671164 on Build GRCh38
rs199779869 Chr.12: 50671174-50671174 on Build GRCh38
rs761173844 Chr.12: 50671146-50671146 on Build GRCh38
rs759889425 Chr.12: 50671178-50671197 on Build GRCh38
rs749601120 Chr.12: 50671189-50671189 on Build GRCh38
rs771662375 Chr.12: 50671213-50671213 on Build GRCh38
rs375668286 Chr.12: 50671206-50671206 on Build GRCh38
rs769160779 Chr.12: 50671195-50671195 on Build GRCh38
rs772759553 Chr.12: 50671214-50671214 on Build GRCh38
rs760132197 Chr.12: 50671220-50671220 on Build GRCh38
rs766235047 Chr.12: 50671222-50671222 on Build GRCh38
rs753589106 Chr.12: 50671156-50671156 on Build GRCh38
rs545686712 Chr.12: 50671150-50671150 on Build GRCh38
rs182272353 Chr.12: 50671153-50671153 on Build GRCh38
rs368139462 Chr.12: 50671166-50671166 on Build GRCh38
rs757476741 Chr.12: 50671240-50671240 on Build GRCh38
rs1034471068 Chr.12: 50671097-50671097 on Build GRCh38

View Full Product Details