Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33360278-33360539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK5RAP1
Entrez Gene ID
51654
Gene Name
CDK5 regulatory subunit associated protein 1
Gene Aliases
C20orf34, C42, CGI-05, HSPC167
UniGene
Hs.435952
Gene Chromosome Location
Chr.20: 33358839-33402768 on Build GRCh38
Transcripts
NM_001278167.1 NM_001278168.1 NM_001278169.1 NM_016082.4 NM_016408.3 XM_011528856.2 XM_011528857.2 XM_011528858.1 XM_011528859.2 XM_011528860.1 XM_017027876.1 XM_017027877.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7084430 c.1575C>A p.G525G Chr.20: 33360459-33360459 on Build GRCh38
5799584 c.1669T>A p.Y557N Chr.20: 33360365-33360365 on Build GRCh38
7086047 c.1620G>A p.M540I Chr.20: 33360414-33360414 on Build GRCh38
310009 c.1624G>T p.D542Y Chr.20: 33360410-33360410 on Build GRCh38
7120356 c.1615G>C p.E539Q Chr.20: 33360419-33360419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749114803 Chr.20: 33360430-33360430 on Build GRCh38
rs755336097 Chr.20: 33360350-33360350 on Build GRCh38
rs766853830 Chr.20: 33360335-33360335 on Build GRCh38
rs369620313 Chr.20: 33360359-33360359 on Build GRCh38
rs989316323 Chr.20: 33360489-33360489 on Build GRCh38
rs200090708 Chr.20: 33360322-33360322 on Build GRCh38
rs956179182 Chr.20: 33360488-33360488 on Build GRCh38
rs544294285 Chr.20: 33360481-33360481 on Build GRCh38
rs753021952 Chr.20: 33360468-33360468 on Build GRCh38
rs781458298 Chr.20: 33360504-33360504 on Build GRCh38
rs201949576 Chr.20: 33360378-33360378 on Build GRCh38
rs767524921 Chr.20: 33360457-33360457 on Build GRCh38
rs200561034 Chr.20: 33360456-33360456 on Build GRCh38
rs557614184 Chr.20: 33360372-33360372 on Build GRCh38
rs561736655 Chr.20: 33360384-33360384 on Build GRCh38
rs368981818 Chr.20: 33360320-33360320 on Build GRCh38
rs762731437 Chr.20: 33360309-33360309 on Build GRCh38
rs769676575 Chr.20: 33360399-33360399 on Build GRCh38
rs750360540 Chr.20: 33360506-33360506 on Build GRCh38
rs77370477 Chr.20: 33360455-33360455 on Build GRCh38
rs765757427 Chr.20: 33360467-33360467 on Build GRCh38
rs545633338 Chr.20: 33360346-33360346 on Build GRCh38
rs761235465 Chr.20: 33360331-33360331 on Build GRCh38
rs572828202 Chr.20: 33360375-33360375 on Build GRCh38
rs970247691 Chr.20: 33360391-33360391 on Build GRCh38
rs1037154402 Chr.20: 33360421-33360421 on Build GRCh38
rs751785823 Chr.20: 33360482-33360482 on Build GRCh38
rs757353978 Chr.20: 33360491-33360491 on Build GRCh38
rs768311774 Chr.20: 33360435-33360435 on Build GRCh38
rs528937618 Chr.20: 33360465-33360465 on Build GRCh38
rs745867001 Chr.20: 33360397-33360397 on Build GRCh38
rs758598698 Chr.20: 33360374-33360374 on Build GRCh38
rs984331508 Chr.20: 33360312-33360312 on Build GRCh38
rs775471148 Chr.20: 33360427-33360427 on Build GRCh38
rs142084089 Chr.20: 33360380-33360380 on Build GRCh38

View Full Product Details