Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32451736-32452007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
UniGene
Hs.585464
Gene Chromosome Location
Chr.10: 32446082-32882864 on Build GRCh38
Transcripts
NM_001026383.2 NM_001321115.1 NM_145023.5 NR_109826.1 NR_109827.1 XM_006717505.1 XM_006717506.1 XM_011519663.1 XM_011519664.1 XM_011519666.1 XM_011519667.1 XM_011519668.1 XM_011519669.1 XM_011519670.1 XM_011519671.1 XM_011519672.1 XM_011519673.1 XM_011519674.1 XM_011519675.1 XM_011519676.1 XM_011519677.1 XM_011519678.1 XM_011519679.1 XM_011519684.2 XM_011519686.2 XM_011519688.2 XM_017016638.1 XM_017016639.1 XM_017016640.1 XM_017016644.1 XM_017016647.1 XM_017016650.1 XM_017016651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5370578 c.157C>T p.L53L Chr.10: 32451799-32451799 on Build GRCh38
4395668 c.252C>T p.I84I Chr.10: 32451894-32451894 on Build GRCh38
1603414 c.109_110insA p.H39fs*2 Chr.10: 32451751-32451752 on Build GRCh38
5370577 c.99T>C p.P33P Chr.10: 32451741-32451741 on Build GRCh38
7307599 c.221C>A p.T74K Chr.10: 32451863-32451863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770776692 Chr.10: 32451807-32451807 on Build GRCh38
rs1022171218 Chr.10: 32451928-32451928 on Build GRCh38
rs977117855 Chr.10: 32451778-32451778 on Build GRCh38
rs143652718 Chr.10: 32451837-32451837 on Build GRCh38
rs746062203 Chr.10: 32451891-32451891 on Build GRCh38
rs752321742 Chr.10: 32451823-32451828 on Build GRCh38
rs967163047 Chr.10: 32451931-32451931 on Build GRCh38
rs754161819 Chr.10: 32451852-32451852 on Build GRCh38
rs554451471 Chr.10: 32451895-32451895 on Build GRCh38
rs1008455494 Chr.10: 32451883-32451883 on Build GRCh38
rs769054239 Chr.10: 32451834-32451834 on Build GRCh38
rs146679927 Chr.10: 32451870-32451871 on Build GRCh38
rs758570771 Chr.10: 32451894-32451894 on Build GRCh38
rs375863955 Chr.10: 32451846-32451846 on Build GRCh38
rs371529800 Chr.10: 32451959-32451959 on Build GRCh38
rs7094838 Chr.10: 32451977-32451977 on Build GRCh38
rs999937582 Chr.10: 32451793-32451793 on Build GRCh38
rs368403420 Chr.10: 32451833-32451833 on Build GRCh38
rs938614566 Chr.10: 32451825-32451825 on Build GRCh38
rs74740427 Chr.10: 32451976-32451976 on Build GRCh38
rs764011965 Chr.10: 32451876-32451876 on Build GRCh38
rs772155435 Chr.10: 32451815-32451815 on Build GRCh38
rs756032098 Chr.10: 32451791-32451791 on Build GRCh38
rs955825496 Chr.10: 32451826-32451826 on Build GRCh38
rs775462645 Chr.10: 32451821-32451821 on Build GRCh38
rs1018467482 Chr.10: 32451885-32451885 on Build GRCh38
rs780608550 Chr.10: 32451780-32451780 on Build GRCh38
rs754489205 Chr.10: 32451776-32451776 on Build GRCh38
rs751315907 Chr.10: 32451769-32451769 on Build GRCh38
rs977162426 Chr.10: 32451955-32451955 on Build GRCh38
rs918630963 Chr.10: 32451889-32451889 on Build GRCh38
rs374470371 Chr.10: 32451790-32451790 on Build GRCh38
rs1037911337 Chr.10: 32451839-32451839 on Build GRCh38
rs759174322 Chr.10: 32451971-32451971 on Build GRCh38
rs757020037 Chr.10: 32451886-32451886 on Build GRCh38
rs768917471 Chr.10: 32451911-32451911 on Build GRCh38
rs748706820 Chr.10: 32451945-32451945 on Build GRCh38
rs746134603 Chr.10: 32451810-32451810 on Build GRCh38
rs1055718131 Chr.10: 32451832-32451832 on Build GRCh38
rs891121473 Chr.10: 32451881-32451881 on Build GRCh38
rs747062894 Chr.10: 32451903-32451903 on Build GRCh38
rs770244521 Chr.10: 32451953-32451953 on Build GRCh38
rs534703724 Chr.10: 32451847-32451847 on Build GRCh38
rs142000877 Chr.10: 32451922-32451922 on Build GRCh38
rs148093587 Chr.10: 32451863-32451863 on Build GRCh38
rs774116810 Chr.10: 32451859-32451859 on Build GRCh38
rs779091860 Chr.10: 32451808-32451808 on Build GRCh38
rs564601002 Chr.10: 32451864-32451864 on Build GRCh38
rs1056842168 Chr.10: 32451938-32451938 on Build GRCh38
rs753827414 Chr.10: 32451882-32451882 on Build GRCh38
rs148965260 Chr.10: 32451794-32451794 on Build GRCh38
rs778715994 Chr.10: 32451890-32451890 on Build GRCh38
rs749089140 Chr.10: 32451801-32451801 on Build GRCh38
rs781553535 Chr.10: 32451915-32451916 on Build GRCh38
rs954423198 Chr.10: 32451854-32451854 on Build GRCh38

View Full Product Details