Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185459166-185459439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC110
Entrez Gene ID
256309
Gene Name
coiled-coil domain containing 110
Gene Aliases
CT52, KM-HN-1, KMHN1
UniGene
Hs.41101
Gene Chromosome Location
Chr.4: 185445182-185471759 on Build GRCh38
Transcripts
NM_001145411.1 NM_152775.3 XM_005262891.3 XM_006714172.2 XM_011531827.2 XM_011531828.2 XM_011531829.2 XM_017007987.1 XM_017007988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7081221 c.1153C>A p.L385I Chr.4: 185459434-185459434 on Build GRCh38
4123744 c.1267C>T p.Q423* Chr.4: 185459320-185459320 on Build GRCh38
5663458 c.1259T>C p.V420A Chr.4: 185459328-185459328 on Build GRCh38
3602477 c.1155C>T p.L385L Chr.4: 185459432-185459432 on Build GRCh38
1428900 c.1332delA p.V445fs*1 Chr.4: 185459255-185459255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145767821 Chr.4: 185459272-185459272 on Build GRCh38
rs766295487 Chr.4: 185459402-185459403 on Build GRCh38
rs765853084 Chr.4: 185459372-185459372 on Build GRCh38
rs113136065 Chr.4: 185459239-185459239 on Build GRCh38
rs773172029 Chr.4: 185459355-185459355 on Build GRCh38
rs757958063 Chr.4: 185459225-185459225 on Build GRCh38
rs779273524 Chr.4: 185459396-185459396 on Build GRCh38
rs768084419 Chr.4: 185459198-185459198 on Build GRCh38
rs764408391 Chr.4: 185459204-185459204 on Build GRCh38
rs570920474 Chr.4: 185459329-185459329 on Build GRCh38
rs771287068 Chr.4: 185459290-185459290 on Build GRCh38
rs941517651 Chr.4: 185459371-185459371 on Build GRCh38
rs753202808 Chr.4: 185459200-185459200 on Build GRCh38
rs780874507 Chr.4: 185459273-185459273 on Build GRCh38
rs780971164 Chr.4: 185459408-185459412 on Build GRCh38
rs146615557 Chr.4: 185459353-185459353 on Build GRCh38
rs754906283 Chr.4: 185459384-185459384 on Build GRCh38
rs769693618 Chr.4: 185459275-185459287 on Build GRCh38
rs756932221 Chr.4: 185459212-185459212 on Build GRCh38
rs974455251 Chr.4: 185459343-185459343 on Build GRCh38
rs749948631 Chr.4: 185459224-185459224 on Build GRCh38
rs572355213 Chr.4: 185459194-185459194 on Build GRCh38
rs565208809 Chr.4: 185459396-185459399 on Build GRCh38
rs764660533 Chr.4: 185459220-185459220 on Build GRCh38
rs753439209 Chr.4: 185459206-185459206 on Build GRCh38
rs145820853 Chr.4: 185459323-185459323 on Build GRCh38
rs761099550 Chr.4: 185459202-185459202 on Build GRCh38
rs751468015 Chr.4: 185459373-185459373 on Build GRCh38
rs369695651 Chr.4: 185459389-185459389 on Build GRCh38
rs971248420 Chr.4: 185459203-185459203 on Build GRCh38
rs746956904 Chr.4: 185459266-185459266 on Build GRCh38
rs772110001 Chr.4: 185459321-185459321 on Build GRCh38
rs749482938 Chr.4: 185459279-185459279 on Build GRCh38
rs11132306 Chr.4: 185459361-185459361 on Build GRCh38
rs781072086 Chr.4: 185459385-185459385 on Build GRCh38
rs756032514 Chr.4: 185459391-185459391 on Build GRCh38
rs748246044 Chr.4: 185459260-185459261 on Build GRCh38
rs764852678 Chr.4: 185459360-185459360 on Build GRCh38
rs776982244 Chr.4: 185459330-185459330 on Build GRCh38
rs144243929 Chr.4: 185459362-185459362 on Build GRCh38
rs778845145 Chr.4: 185459305-185459305 on Build GRCh38
rs114701079 Chr.4: 185459301-185459301 on Build GRCh38
rs927937001 Chr.4: 185459304-185459304 on Build GRCh38
rs112643870 Chr.4: 185459325-185459325 on Build GRCh38

View Full Product Details