Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60874553-60874814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
UniGene
Hs.655028
Gene Chromosome Location
Chr.17: 60677811-61392838 on Build GRCh38
Transcripts
NM_001099432.1 NM_001320470.1 NM_017679.3 XM_005257472.1 XM_011524939.1 XM_011524940.1 XM_011524941.1 XM_011524942.2 XM_011524943.2 XM_011524944.1 XM_011524945.2 XM_011524946.2 XM_011524947.1 XM_011524948.2 XM_011524949.2 XM_011524950.1 XM_011524951.2 XM_017024783.1 XM_017024784.1 XM_017024785.1 XM_017024786.1 XM_017024787.1 XM_017024788.1 XM_017024789.1 XM_017024790.1 XM_017024791.1 XM_017024792.1 XM_017024793.1 XM_017024794.1 XM_017024795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417416 c.641C>T p.T214M Chr.17: 60874718-60874718 on Build GRCh38
3520365 c.651C>T p.F217F Chr.17: 60874728-60874728 on Build GRCh38
5668055 c.607G>A p.E203K Chr.17: 60874684-60874684 on Build GRCh38
3795892 c.641C>T p.T214M Chr.17: 60874718-60874718 on Build GRCh38
3421749 c.642G>A p.T214T Chr.17: 60874719-60874719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371725896 Chr.17: 60874618-60874618 on Build GRCh38
rs768203819 Chr.17: 60874785-60874785 on Build GRCh38
rs186183906 Chr.17: 60874775-60874775 on Build GRCh38
rs745854936 Chr.17: 60874759-60874759 on Build GRCh38
rs779916875 Chr.17: 60874638-60874638 on Build GRCh38
rs753617921 Chr.17: 60874749-60874749 on Build GRCh38
rs151089810 Chr.17: 60874622-60874622 on Build GRCh38
rs751009642 Chr.17: 60874672-60874672 on Build GRCh38
rs760266975 Chr.17: 60874644-60874644 on Build GRCh38
rs941223043 Chr.17: 60874774-60874774 on Build GRCh38
rs765823513 Chr.17: 60874671-60874671 on Build GRCh38
rs771608528 Chr.17: 60874635-60874635 on Build GRCh38
rs776305673 Chr.17: 60874760-60874762 on Build GRCh38
rs777567245 Chr.17: 60874741-60874741 on Build GRCh38
rs114101440 Chr.17: 60874620-60874620 on Build GRCh38
rs746483225 Chr.17: 60874645-60874646 on Build GRCh38
rs889550428 Chr.17: 60874780-60874780 on Build GRCh38
rs938629674 Chr.17: 60874642-60874642 on Build GRCh38
rs371881802 Chr.17: 60874648-60874648 on Build GRCh38
rs771562337 Chr.17: 60874767-60874767 on Build GRCh38
rs768333656 Chr.17: 60874750-60874750 on Build GRCh38
rs1021091649 Chr.17: 60874583-60874583 on Build GRCh38
rs757969491 Chr.17: 60874624-60874624 on Build GRCh38
rs1024461843 Chr.17: 60874653-60874653 on Build GRCh38
rs746602205 Chr.17: 60874778-60874778 on Build GRCh38
rs751769056 Chr.17: 60874719-60874719 on Build GRCh38
rs762349606 Chr.17: 60874669-60874669 on Build GRCh38
rs775185589 Chr.17: 60874643-60874643 on Build GRCh38
rs375377185 Chr.17: 60874725-60874725 on Build GRCh38
rs749777847 Chr.17: 60874784-60874787 on Build GRCh38
rs369969114 Chr.17: 60874751-60874751 on Build GRCh38
rs763826015 Chr.17: 60874645-60874645 on Build GRCh38
rs540900531 Chr.17: 60874589-60874589 on Build GRCh38
rs766394914 Chr.17: 60874718-60874718 on Build GRCh38
rs776860229 Chr.17: 60874651-60874651 on Build GRCh38

View Full Product Details