Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 15936284-15936547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL7
Entrez Gene ID
23194
Gene Name
F-box and leucine rich repeat protein 7
Gene Aliases
FBL6, FBL7
UniGene
Hs.433057
Gene Chromosome Location
Chr.5: 15500196-15939796 on Build GRCh38
Transcripts
NM_001278317.1 NM_012304.4 XM_011513998.1 XM_017009262.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5719064 c.805G>A p.G269S Chr.5: 15936515-15936515 on Build GRCh38
332067 c.822C>T p.I274I Chr.5: 15936532-15936532 on Build GRCh38
5424211 c.768G>C p.L256F Chr.5: 15936478-15936478 on Build GRCh38
6103155 c.746C>G p.S249C Chr.5: 15936456-15936456 on Build GRCh38
6170373 c.773G>T p.R258L Chr.5: 15936483-15936483 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144246465 Chr.5: 15936313-15936313 on Build GRCh38
rs772815727 Chr.5: 15936476-15936476 on Build GRCh38
rs776423724 Chr.5: 15936459-15936459 on Build GRCh38
rs748935681 Chr.5: 15936441-15936442 on Build GRCh38
rs576217444 Chr.5: 15936502-15936502 on Build GRCh38
rs1026279759 Chr.5: 15936387-15936387 on Build GRCh38
rs568110410 Chr.5: 15936333-15936333 on Build GRCh38
rs754453330 Chr.5: 15936514-15936514 on Build GRCh38
rs780230488 Chr.5: 15936488-15936488 on Build GRCh38
rs1042573419 Chr.5: 15936518-15936518 on Build GRCh38
rs370550112 Chr.5: 15936474-15936474 on Build GRCh38
rs374393167 Chr.5: 15936415-15936415 on Build GRCh38
rs535498084 Chr.5: 15936391-15936391 on Build GRCh38
rs993614596 Chr.5: 15936363-15936363 on Build GRCh38
rs571441703 Chr.5: 15936401-15936401 on Build GRCh38
rs954876547 Chr.5: 15936389-15936389 on Build GRCh38
rs899246148 Chr.5: 15936357-15936357 on Build GRCh38
rs557631491 Chr.5: 15936479-15936479 on Build GRCh38
rs189208873 Chr.5: 15936483-15936483 on Build GRCh38
rs1016129588 Chr.5: 15936330-15936330 on Build GRCh38
rs965059043 Chr.5: 15936365-15936365 on Build GRCh38
rs764521002 Chr.5: 15936511-15936511 on Build GRCh38
rs549432925 Chr.5: 15936409-15936409 on Build GRCh38
rs148473239 Chr.5: 15936460-15936460 on Build GRCh38
rs565687593 Chr.5: 15936366-15936366 on Build GRCh38
rs908718491 Chr.5: 15936338-15936338 on Build GRCh38
rs1053473569 Chr.5: 15936464-15936464 on Build GRCh38
rs750542921 Chr.5: 15936411-15936411 on Build GRCh38
rs746917192 Chr.5: 15936446-15936446 on Build GRCh38
rs761175476 Chr.5: 15936490-15936490 on Build GRCh38
rs780048506 Chr.5: 15936443-15936443 on Build GRCh38
rs746159716 Chr.5: 15936420-15936420 on Build GRCh38
rs553757608 Chr.5: 15936402-15936402 on Build GRCh38
rs758521502 Chr.5: 15936422-15936422 on Build GRCh38
rs779263460 Chr.5: 15936414-15936414 on Build GRCh38

View Full Product Details