Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22043122-22043381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF17
Entrez Gene ID
8822
Gene Name
fibroblast growth factor 17
Gene Aliases
FGF-13, FGF-17, HH20
UniGene
Hs.248192
Gene Chromosome Location
Chr.8: 22039708-22048809 on Build GRCh38
Transcripts
NM_001304478.1 NM_003867.3 XM_011544683.1 XM_011544684.2 XM_011544685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6663515 c.57C>G p.L19L Chr.8: 22043166-22043166 on Build GRCh38
6663516 c.55C>A p.L19I Chr.8: 22043164-22043164 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs190030914 Chr.8: 22043192-22043192 on Build GRCh38
rs577326455 Chr.8: 22043198-22043198 on Build GRCh38
rs551472660 Chr.8: 22043329-22043329 on Build GRCh38
rs192575252 Chr.8: 22043350-22043350 on Build GRCh38
rs911560313 Chr.8: 22043243-22043243 on Build GRCh38
rs767432143 Chr.8: 22043268-22043268 on Build GRCh38
rs368478275 Chr.8: 22043200-22043200 on Build GRCh38
rs986957591 Chr.8: 22043346-22043346 on Build GRCh38
rs3176271 Chr.8: 22043238-22043238 on Build GRCh38
rs750014801 Chr.8: 22043142-22043142 on Build GRCh38
rs761428899 Chr.8: 22043221-22043221 on Build GRCh38
rs149043820 Chr.8: 22043188-22043188 on Build GRCh38
rs545540109 Chr.8: 22043158-22043158 on Build GRCh38
rs900779 Chr.8: 22043246-22043246 on Build GRCh38
rs758266120 Chr.8: 22043147-22043147 on Build GRCh38
rs764633548 Chr.8: 22043223-22043223 on Build GRCh38
rs759910488 Chr.8: 22043204-22043204 on Build GRCh38
rs969798145 Chr.8: 22043318-22043318 on Build GRCh38
rs762510668 Chr.8: 22043227-22043227 on Build GRCh38
rs751566846 Chr.8: 22043171-22043171 on Build GRCh38
rs933466631 Chr.8: 22043340-22043340 on Build GRCh38
rs369581023 Chr.8: 22043247-22043247 on Build GRCh38
rs946564822 Chr.8: 22043286-22043286 on Build GRCh38
rs372106895 Chr.8: 22043228-22043228 on Build GRCh38
rs367564532 Chr.8: 22043224-22043224 on Build GRCh38
rs374293819 Chr.8: 22043202-22043202 on Build GRCh38
rs188295078 Chr.8: 22043339-22043339 on Build GRCh38
rs999396952 Chr.8: 22043239-22043239 on Build GRCh38
rs972229378 Chr.8: 22043332-22043332 on Build GRCh38
rs772732594 Chr.8: 22043207-22043207 on Build GRCh38
rs1016963067 Chr.8: 22043326-22043326 on Build GRCh38
rs764977035 Chr.8: 22043141-22043141 on Build GRCh38
rs754914249 Chr.8: 22043175-22043175 on Build GRCh38
rs984775671 Chr.8: 22043321-22043321 on Build GRCh38
rs546244986 Chr.8: 22043253-22043253 on Build GRCh38
rs776104827 Chr.8: 22043209-22043209 on Build GRCh38
rs942393326 Chr.8: 22043358-22043358 on Build GRCh38
rs200967245 Chr.8: 22043194-22043194 on Build GRCh38
rs370876306 Chr.8: 22043187-22043187 on Build GRCh38
rs1023698817 Chr.8: 22043272-22043272 on Build GRCh38
rs182099999 Chr.8: 22043252-22043252 on Build GRCh38
rs200097089 Chr.8: 22043201-22043201 on Build GRCh38
rs751369578 Chr.8: 22043232-22043232 on Build GRCh38

View Full Product Details