Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122836842-122837073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUZD1
Entrez Gene ID
50624
Gene Name
CUB and zona pellucida like domains 1
Gene Aliases
ERG-1, ERG1, ITMAP1, UO-44
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122846175 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Gene Symbol
FAM24B-CUZD1
Entrez Gene ID
100533195
Gene Name
FAM24B-CUZD1 readthrough
Gene Aliases
-
UniGene
Hs.647182
Gene Chromosome Location
Chr.10: 122832155-122879641 on Build GRCh38
Transcripts
NM_022034.5 NR_037912.1 NR_037915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
107330 c.713C>T p.S238L Chr.10: 122836935-122836935 on Build GRCh38
3670478 c.766C>A p.R256R Chr.10: 122836882-122836882 on Build GRCh38
5359881 c.600-1G>A p.? Chr.10: 122837049-122837049 on Build GRCh38
915718 c.708C>T p.F236F Chr.10: 122836940-122836940 on Build GRCh38
915717 c.776C>T p.S259F Chr.10: 122836872-122836872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201090823 Chr.10: 122836899-122836899 on Build GRCh38
rs779064067 Chr.10: 122837029-122837029 on Build GRCh38
rs749658016 Chr.10: 122837022-122837022 on Build GRCh38
rs182334751 Chr.10: 122836881-122836881 on Build GRCh38
rs148175429 Chr.10: 122836994-122836994 on Build GRCh38
rs775893228 Chr.10: 122836908-122836908 on Build GRCh38
rs373493047 Chr.10: 122836910-122836910 on Build GRCh38
rs781778473 Chr.10: 122836916-122836916 on Build GRCh38
rs543599364 Chr.10: 122837014-122837014 on Build GRCh38
rs768438076 Chr.10: 122837052-122837052 on Build GRCh38
rs756644803 Chr.10: 122837004-122837004 on Build GRCh38
rs752424627 Chr.10: 122836892-122836892 on Build GRCh38
rs200891112 Chr.10: 122836967-122836967 on Build GRCh38
rs748311677 Chr.10: 122837030-122837030 on Build GRCh38
rs761933837 Chr.10: 122836874-122836874 on Build GRCh38
rs762775582 Chr.10: 122836974-122836974 on Build GRCh38
rs763423390 Chr.10: 122836945-122836946 on Build GRCh38
rs763718858 Chr.10: 122836980-122836980 on Build GRCh38
rs763778389 Chr.10: 122836898-122836898 on Build GRCh38
rs144483251 Chr.10: 122836956-122836956 on Build GRCh38
rs767065614 Chr.10: 122836995-122836995 on Build GRCh38
rs768812676 Chr.10: 122836940-122836940 on Build GRCh38
rs774604884 Chr.10: 122836946-122836946 on Build GRCh38
rs775198104 Chr.10: 122836973-122836973 on Build GRCh38
rs755366577 Chr.10: 122837025-122837025 on Build GRCh38
rs751208431 Chr.10: 122836992-122836992 on Build GRCh38
rs751069867 Chr.10: 122836911-122836911 on Build GRCh38
rs748244519 Chr.10: 122836950-122836950 on Build GRCh38
rs973568574 Chr.10: 122837034-122837034 on Build GRCh38
rs138928505 Chr.10: 122836913-122836913 on Build GRCh38
rs747068473 Chr.10: 122837038-122837038 on Build GRCh38
rs148411775 Chr.10: 122837040-122837040 on Build GRCh38
rs943994852 Chr.10: 122836905-122836905 on Build GRCh38
rs776548188 Chr.10: 122836879-122836879 on Build GRCh38
rs558489122 Chr.10: 122836882-122836882 on Build GRCh38
rs766345318 Chr.10: 122836878-122836878 on Build GRCh38
rs369603738 Chr.10: 122836939-122836939 on Build GRCh38
rs201852136 Chr.10: 122836954-122836954 on Build GRCh38
rs762712325 Chr.10: 122837041-122837041 on Build GRCh38
rs143377209 Chr.10: 122836935-122836935 on Build GRCh38
rs765037678 Chr.10: 122836972-122836972 on Build GRCh38
rs751001597 Chr.10: 122837003-122837003 on Build GRCh38
rs773874915 Chr.10: 122837054-122837054 on Build GRCh38
rs117062933 Chr.10: 122836912-122836912 on Build GRCh38
rs757938786 Chr.10: 122836895-122836895 on Build GRCh38
rs867748457 Chr.10: 122837000-122837000 on Build GRCh38
rs890450802 Chr.10: 122836886-122836886 on Build GRCh38
rs149276651 Chr.10: 122836919-122836919 on Build GRCh38
rs201913985 Chr.10: 122836934-122836934 on Build GRCh38
rs561395165 Chr.10: 122837033-122837033 on Build GRCh38
rs373241545 Chr.10: 122837036-122837036 on Build GRCh38

View Full Product Details