Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241503901-241504132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6925318 c.1036G>T p.G346C Chr.1: 241504114-241504114 on Build GRCh38
679716 c.1052C>A p.S351* Chr.1: 241504098-241504098 on Build GRCh38
4684647 c.1051T>A p.S351T Chr.1: 241504099-241504099 on Build GRCh38
6967256 c.1018A>G p.N340D Chr.1: 241504132-241504132 on Build GRCh38
4768017 c.1108+6C>T p.? Chr.1: 241504036-241504036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200118475 Chr.1: 241504039-241504039 on Build GRCh38
rs951913314 Chr.1: 241504082-241504082 on Build GRCh38
rs863223966 Chr.1: 241504057-241504057 on Build GRCh38
rs1040420520 Chr.1: 241503985-241503985 on Build GRCh38
rs1027877625 Chr.1: 241504081-241504081 on Build GRCh38
rs959164482 Chr.1: 241503943-241503943 on Build GRCh38
rs555908729 Chr.1: 241503978-241503978 on Build GRCh38
rs863223987 Chr.1: 241504044-241504046 on Build GRCh38
rs775548838 Chr.1: 241503996-241503996 on Build GRCh38
rs918902822 Chr.1: 241504078-241504078 on Build GRCh38
rs201513242 Chr.1: 241504032-241504032 on Build GRCh38
rs121913119 Chr.1: 241504066-241504066 on Build GRCh38
rs4660099 Chr.1: 241503933-241503933 on Build GRCh38
rs565940535 Chr.1: 241503927-241503927 on Build GRCh38
rs756469140 Chr.1: 241504064-241504067 on Build GRCh38
rs4659607 Chr.1: 241503944-241503944 on Build GRCh38
rs758440879 Chr.1: 241504094-241504094 on Build GRCh38
rs863224004 Chr.1: 241504053-241504053 on Build GRCh38
rs200550559 Chr.1: 241504013-241504013 on Build GRCh38
rs190353292 Chr.1: 241503945-241503945 on Build GRCh38
rs863224016 Chr.1: 241504093-241504094 on Build GRCh38
rs539449056 Chr.1: 241503946-241503946 on Build GRCh38
rs576072868 Chr.1: 241504029-241504029 on Build GRCh38
rs761954764 Chr.1: 241504033-241504033 on Build GRCh38
rs1008895929 Chr.1: 241503984-241503984 on Build GRCh38
rs946935917 Chr.1: 241503924-241503924 on Build GRCh38
rs766918221 Chr.1: 241504099-241504099 on Build GRCh38
rs749316923 Chr.1: 241504101-241504101 on Build GRCh38
rs750535216 Chr.1: 241504073-241504073 on Build GRCh38
rs727503927 Chr.1: 241504083-241504083 on Build GRCh38
rs772180108 Chr.1: 241503992-241503992 on Build GRCh38
rs765397179 Chr.1: 241504035-241504035 on Build GRCh38
rs368482304 Chr.1: 241504011-241504011 on Build GRCh38
rs1057517734 Chr.1: 241504041-241504041 on Build GRCh38
rs755436052 Chr.1: 241504102-241504102 on Build GRCh38
rs760580659 Chr.1: 241504004-241504004 on Build GRCh38

View Full Product Details