Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49511896-49512153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYATL3
Entrez Gene ID
389396
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
C6orf140, bA28H17.2
UniGene
Hs.585172
Gene Chromosome Location
Chr.6: 49499958-49528064 on Build GRCh38
Transcripts
NM_001010904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3949302 c.78+4C>G p.? Chr.6: 49512072-49512072 on Build GRCh38
3949301 c.64C>A p.P22T Chr.6: 49512054-49512054 on Build GRCh38
7251689 c.75C>A p.L25L Chr.6: 49512065-49512065 on Build GRCh38
1137536 c.47T>C p.M16T Chr.6: 49512037-49512037 on Build GRCh38
3762114 c.72A>T p.S24S Chr.6: 49512062-49512062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913991099 Chr.6: 49511955-49511955 on Build GRCh38
rs986063674 Chr.6: 49512121-49512121 on Build GRCh38
rs141201277 Chr.6: 49511981-49511981 on Build GRCh38
rs1034722524 Chr.6: 49511967-49511967 on Build GRCh38
rs1480615 Chr.6: 49512062-49512062 on Build GRCh38
rs776917224 Chr.6: 49511968-49511968 on Build GRCh38
rs1057138792 Chr.6: 49512032-49512032 on Build GRCh38
rs954749618 Chr.6: 49512090-49512090 on Build GRCh38
rs546529523 Chr.6: 49512043-49512043 on Build GRCh38
rs36125184 Chr.6: 49512053-49512054 on Build GRCh38
rs9473579 Chr.6: 49511935-49511935 on Build GRCh38
rs560399915 Chr.6: 49512028-49512028 on Build GRCh38
rs905191475 Chr.6: 49512007-49512007 on Build GRCh38
rs760307636 Chr.6: 49512105-49512105 on Build GRCh38
rs534613072 Chr.6: 49512038-49512038 on Build GRCh38
rs759132418 Chr.6: 49512046-49512046 on Build GRCh38
rs1043742293 Chr.6: 49512000-49512000 on Build GRCh38
rs557104763 Chr.6: 49512087-49512087 on Build GRCh38
rs1021703069 Chr.6: 49512034-49512034 on Build GRCh38
rs13193160 Chr.6: 49512108-49512108 on Build GRCh38
rs538484418 Chr.6: 49512085-49512085 on Build GRCh38
rs13193063 Chr.6: 49512037-49512037 on Build GRCh38

View Full Product Details