Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10137150-10137377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT1
Entrez Gene ID
1786
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
UniGene
Hs.202672
Gene Chromosome Location
Chr.19: 10133344-10195135 on Build GRCh38
Transcripts
NM_001130823.2 NM_001318730.1 NM_001318731.1 NM_001379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3528092 c.4246-1G>A p.? Chr.19: 10137281-10137281 on Build GRCh38
6948302 c.4255G>A p.A1419T Chr.19: 10137271-10137271 on Build GRCh38
990435 c.4358G>T p.R1453M Chr.19: 10137168-10137168 on Build GRCh38
6647738 c.4311C>T p.R1437R Chr.19: 10137215-10137215 on Build GRCh38
5388532 c.4294C>T p.P1432S Chr.19: 10137232-10137232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750474625 Chr.19: 10137298-10137298 on Build GRCh38
rs527614200 Chr.19: 10137324-10137324 on Build GRCh38
rs200416276 Chr.19: 10137182-10137182 on Build GRCh38
rs769675693 Chr.19: 10137193-10137193 on Build GRCh38
rs542862981 Chr.19: 10137336-10137336 on Build GRCh38
rs762360656 Chr.19: 10137283-10137283 on Build GRCh38
rs35012154 Chr.19: 10137353-10137354 on Build GRCh38
rs757066879 Chr.19: 10137296-10137297 on Build GRCh38
rs748089711 Chr.19: 10137192-10137192 on Build GRCh38
rs1000743307 Chr.19: 10137346-10137346 on Build GRCh38
rs555878888 Chr.19: 10137258-10137258 on Build GRCh38
rs754833277 Chr.19: 10137181-10137181 on Build GRCh38
rs2229858 Chr.19: 10137206-10137206 on Build GRCh38
rs749134980 Chr.19: 10137242-10137242 on Build GRCh38
rs909222539 Chr.19: 10137294-10137294 on Build GRCh38
rs1053592193 Chr.19: 10137355-10137355 on Build GRCh38
rs137949464 Chr.19: 10137310-10137310 on Build GRCh38
rs1031456207 Chr.19: 10137277-10137277 on Build GRCh38
rs139918621 Chr.19: 10137250-10137250 on Build GRCh38
rs755314598 Chr.19: 10137313-10137313 on Build GRCh38
rs774010132 Chr.19: 10137232-10137232 on Build GRCh38
rs2290683 Chr.19: 10137299-10137299 on Build GRCh38
rs770571074 Chr.19: 10137229-10137229 on Build GRCh38
rs79585069 Chr.19: 10137297-10137297 on Build GRCh38
rs752585204 Chr.19: 10137327-10137327 on Build GRCh38
rs577313883 Chr.19: 10137272-10137272 on Build GRCh38
rs748782322 Chr.19: 10137215-10137215 on Build GRCh38
rs1026801149 Chr.19: 10137331-10137331 on Build GRCh38
rs368411249 Chr.19: 10137329-10137329 on Build GRCh38
rs376184130 Chr.19: 10137328-10137328 on Build GRCh38

View Full Product Details