Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78925207-78925464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSH
Entrez Gene ID
1512
Gene Name
cathepsin H
Gene Aliases
ACC-4, ACC-5, ACC4, ACC5, CPSB
UniGene
Hs.148641
Gene Chromosome Location
Chr.15: 78921750-78945098 on Build GRCh38
Transcripts
NM_001319137.1 NM_004390.4 XM_005254181.2 XM_017021951.1 XM_017021952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637810 c.743C>A p.P248H Chr.15: 78925397-78925397 on Build GRCh38
7230158 c.791C>T p.T264I Chr.15: 78925349-78925349 on Build GRCh38
4505903 c.705C>G p.D235E Chr.15: 78925435-78925435 on Build GRCh38
3936864 c.801C>T p.Y267Y Chr.15: 78925339-78925339 on Build GRCh38
6637811 c.806+2T>A p.? Chr.15: 78925332-78925332 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025226771 Chr.15: 78925230-78925230 on Build GRCh38
rs911124601 Chr.15: 78925266-78925266 on Build GRCh38
rs762604293 Chr.15: 78925403-78925403 on Build GRCh38
rs964018399 Chr.15: 78925267-78925267 on Build GRCh38
rs755265082 Chr.15: 78925438-78925438 on Build GRCh38
rs770419158 Chr.15: 78925373-78925373 on Build GRCh38
rs368961648 Chr.15: 78925314-78925314 on Build GRCh38
rs149316890 Chr.15: 78925435-78925435 on Build GRCh38
rs137865366 Chr.15: 78925306-78925306 on Build GRCh38
rs750468764 Chr.15: 78925432-78925432 on Build GRCh38
rs747510233 Chr.15: 78925434-78925434 on Build GRCh38
rs905424000 Chr.15: 78925391-78925391 on Build GRCh38
rs546822356 Chr.15: 78925384-78925384 on Build GRCh38
rs747159161 Chr.15: 78925343-78925343 on Build GRCh38
rs572992158 Chr.15: 78925249-78925249 on Build GRCh38
rs900661408 Chr.15: 78925257-78925257 on Build GRCh38
rs777816322 Chr.15: 78925313-78925313 on Build GRCh38
rs779091243 Chr.15: 78925271-78925271 on Build GRCh38
rs759388714 Chr.15: 78925298-78925298 on Build GRCh38
rs1045117316 Chr.15: 78925385-78925385 on Build GRCh38
rs757254632 Chr.15: 78925344-78925344 on Build GRCh38
rs780573628 Chr.15: 78925357-78925357 on Build GRCh38
rs757471478 Chr.15: 78925426-78925426 on Build GRCh38
rs560698092 Chr.15: 78925427-78925427 on Build GRCh38
rs138844057 Chr.15: 78925362-78925362 on Build GRCh38
rs2289695 Chr.15: 78925289-78925289 on Build GRCh38
rs202111900 Chr.15: 78925433-78925433 on Build GRCh38
rs766349549 Chr.15: 78925286-78925286 on Build GRCh38
rs758649696 Chr.15: 78925307-78925307 on Build GRCh38
rs2289694 Chr.15: 78925264-78925264 on Build GRCh38
rs751728319 Chr.15: 78925419-78925419 on Build GRCh38
rs955039775 Chr.15: 78925246-78925246 on Build GRCh38
rs781426096 Chr.15: 78925342-78925345 on Build GRCh38
rs764998902 Chr.15: 78925300-78925300 on Build GRCh38
rs1006489188 Chr.15: 78925398-78925398 on Build GRCh38
rs1014885271 Chr.15: 78925292-78925292 on Build GRCh38
rs765090887 Chr.15: 78925400-78925400 on Build GRCh38
rs764418306 Chr.15: 78925413-78925413 on Build GRCh38
rs2044226 Chr.15: 78925348-78925348 on Build GRCh38
rs117238468 Chr.15: 78925349-78925349 on Build GRCh38
rs772715662 Chr.15: 78925370-78925370 on Build GRCh38
rs771781236 Chr.15: 78925365-78925365 on Build GRCh38
rs775384459 Chr.15: 78925401-78925401 on Build GRCh38

View Full Product Details