Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 184795385-184795614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM129A
Entrez Gene ID
116496
Gene Name
family with sequence similarity 129 member A
Gene Aliases
C1orf24, GIG39, NIBAN
UniGene
Hs.518662
Gene Chromosome Location
Chr.1: 184791025-184975132 on Build GRCh38
Transcripts
NM_052966.3 XM_011509140.2 XM_011509141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251735 c.2323C>A p.P775T Chr.1: 184795441-184795441 on Build GRCh38
74579 c.2182C>A p.P728T Chr.1: 184795582-184795582 on Build GRCh38
5832393 c.2330delG p.C777fs*15 Chr.1: 184795434-184795434 on Build GRCh38
5844793 c.2360delG p.G787fs*5 Chr.1: 184795404-184795404 on Build GRCh38
5844794 c.2251G>A p.E751K Chr.1: 184795513-184795513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140276880 Chr.1: 184795546-184795546 on Build GRCh38
rs771684949 Chr.1: 184795574-184795574 on Build GRCh38
rs569878628 Chr.1: 184795487-184795487 on Build GRCh38
rs747683734 Chr.1: 184795573-184795573 on Build GRCh38
rs763670581 Chr.1: 184795409-184795409 on Build GRCh38
rs753885053 Chr.1: 184795442-184795442 on Build GRCh38
rs766847624 Chr.1: 184795517-184795517 on Build GRCh38
rs770437163 Chr.1: 184795521-184795523 on Build GRCh38
rs536032437 Chr.1: 184795562-184795562 on Build GRCh38
rs201876014 Chr.1: 184795563-184795563 on Build GRCh38
rs566736052 Chr.1: 184795566-184795566 on Build GRCh38
rs769733220 Chr.1: 184795457-184795457 on Build GRCh38
rs773796065 Chr.1: 184795589-184795590 on Build GRCh38
rs370722883 Chr.1: 184795429-184795429 on Build GRCh38
rs758646793 Chr.1: 184795545-184795545 on Build GRCh38
rs779195910 Chr.1: 184795447-184795447 on Build GRCh38
rs750801506 Chr.1: 184795430-184795430 on Build GRCh38
rs994141230 Chr.1: 184795491-184795491 on Build GRCh38
rs761431817 Chr.1: 184795479-184795479 on Build GRCh38
rs761520308 Chr.1: 184795426-184795426 on Build GRCh38
rs778344193 Chr.1: 184795568-184795568 on Build GRCh38
rs537335051 Chr.1: 184795494-184795494 on Build GRCh38
rs752967343 Chr.1: 184795451-184795451 on Build GRCh38
rs35545276 Chr.1: 184795547-184795547 on Build GRCh38
rs373138315 Chr.1: 184795456-184795456 on Build GRCh38
rs376365064 Chr.1: 184795472-184795472 on Build GRCh38
rs755876221 Chr.1: 184795557-184795557 on Build GRCh38
rs750908430 Chr.1: 184795453-184795453 on Build GRCh38
rs758736883 Chr.1: 184795452-184795452 on Build GRCh38
rs765378611 Chr.1: 184795539-184795539 on Build GRCh38
rs773978173 Chr.1: 184795423-184795423 on Build GRCh38
rs747178668 Chr.1: 184795481-184795481 on Build GRCh38
rs76530866 Chr.1: 184795467-184795467 on Build GRCh38
rs762481558 Chr.1: 184795407-184795407 on Build GRCh38
rs192126521 Chr.1: 184795543-184795543 on Build GRCh38
rs969567506 Chr.1: 184795564-184795564 on Build GRCh38
rs760431201 Chr.1: 184795584-184795584 on Build GRCh38
rs779860728 Chr.1: 184795463-184795463 on Build GRCh38
rs148356793 Chr.1: 184795483-184795483 on Build GRCh38
rs200364804 Chr.1: 184795538-184795538 on Build GRCh38
rs748904031 Chr.1: 184795498-184795498 on Build GRCh38
rs143488738 Chr.1: 184795484-184795484 on Build GRCh38
rs766823696 Chr.1: 184795438-184795438 on Build GRCh38
rs996687440 Chr.1: 184795411-184795411 on Build GRCh38
rs755118661 Chr.1: 184795443-184795443 on Build GRCh38
rs777499981 Chr.1: 184795426-184795427 on Build GRCh38
rs768149242 Chr.1: 184795469-184795469 on Build GRCh38
rs370794770 Chr.1: 184795575-184795575 on Build GRCh38
rs754387667 Chr.1: 184795519-184795519 on Build GRCh38
rs761191983 Chr.1: 184795437-184795437 on Build GRCh38
rs373614339 Chr.1: 184795593-184795593 on Build GRCh38
rs771198252 Chr.1: 184795592-184795592 on Build GRCh38

View Full Product Details