Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171872805-171873078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW11
Entrez Gene ID
23291
Gene Name
F-box and WD repeat domain containing 11
Gene Aliases
BTRC2, BTRCP2, FBW1B, FBXW1B, Fbw11, Hos
UniGene
Hs.484138
Gene Chromosome Location
Chr.5: 171861549-172006873 on Build GRCh38
Transcripts
NM_012300.2 NM_033644.2 NM_033645.2 XM_005265855.4 XM_005265856.2 XM_005265857.2 XM_005265858.2 XM_017009279.1 XM_017009280.1 XM_017009281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5060937 c.1242C>T p.R414R Chr.5: 171872907-171872907 on Build GRCh38
3853703 c.1241G>A p.R414H Chr.5: 171872908-171872908 on Build GRCh38
1132209 c.1170G>T p.T390T Chr.5: 171872979-171872979 on Build GRCh38
4997647 c.1219G>A p.A407T Chr.5: 171872930-171872930 on Build GRCh38
7314186 c.1186C>A p.L396I Chr.5: 171872924-171872924 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151231711 Chr.5: 171872916-171872916 on Build GRCh38
rs746414257 Chr.5: 171872974-171872974 on Build GRCh38
rs551242596 Chr.5: 171873028-171873028 on Build GRCh38
rs773915115 Chr.5: 171873007-171873007 on Build GRCh38
rs770420418 Chr.5: 171872970-171872970 on Build GRCh38
rs201900468 Chr.5: 171873036-171873036 on Build GRCh38
rs774017484 Chr.5: 171873032-171873032 on Build GRCh38
rs760232552 Chr.5: 171873038-171873038 on Build GRCh38
rs369727246 Chr.5: 171873020-171873020 on Build GRCh38
rs780292305 Chr.5: 171872929-171872929 on Build GRCh38
rs375486061 Chr.5: 171872855-171872855 on Build GRCh38
rs763993568 Chr.5: 171873039-171873039 on Build GRCh38
rs747224602 Chr.5: 171872937-171872937 on Build GRCh38
rs748600750 Chr.5: 171872959-171872959 on Build GRCh38
rs140264158 Chr.5: 171872943-171872943 on Build GRCh38
rs199652940 Chr.5: 171872913-171872913 on Build GRCh38
rs756047584 Chr.5: 171872880-171872880 on Build GRCh38
rs752618955 Chr.5: 171872863-171872863 on Build GRCh38
rs975926806 Chr.5: 171872836-171872836 on Build GRCh38
rs779546687 Chr.5: 171873002-171873003 on Build GRCh38
rs759136194 Chr.5: 171873035-171873035 on Build GRCh38
rs753613670 Chr.5: 171872914-171872914 on Build GRCh38
rs755035189 Chr.5: 171872940-171872940 on Build GRCh38
rs766970669 Chr.5: 171872859-171872859 on Build GRCh38
rs771245045 Chr.5: 171873026-171873026 on Build GRCh38

View Full Product Details