Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 73831512-73831761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN4
Entrez Gene ID
1364
Gene Name
claudin 4
Gene Aliases
CPE-R, CPER, CPETR, CPETR1, WBSCR8, hCPE-R
UniGene
Hs.647036
Gene Chromosome Location
Chr.7: 73830863-73832693 on Build GRCh38
Transcripts
NM_001305.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1451999 c.532delG p.L180fs*>30 Chr.7: 73831733-73831733 on Build GRCh38
5528734 c.486C>T p.A162A Chr.7: 73831687-73831687 on Build GRCh38
1091601 c.520C>T p.L174L Chr.7: 73831721-73831721 on Build GRCh38
6885587 c.525C>T p.L175L Chr.7: 73831726-73831726 on Build GRCh38
5407861 c.432C>T p.I144I Chr.7: 73831633-73831633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368820080 Chr.7: 73831646-73831646 on Build GRCh38
rs141788310 Chr.7: 73831696-73831696 on Build GRCh38
rs994139151 Chr.7: 73831665-73831665 on Build GRCh38
rs782815880 Chr.7: 73831552-73831552 on Build GRCh38
rs782441051 Chr.7: 73831672-73831672 on Build GRCh38
rs200176069 Chr.7: 73831689-73831689 on Build GRCh38
rs575127499 Chr.7: 73831587-73831587 on Build GRCh38
rs782701130 Chr.7: 73831715-73831715 on Build GRCh38
rs200778903 Chr.7: 73831656-73831656 on Build GRCh38
rs782014458 Chr.7: 73831709-73831709 on Build GRCh38
rs147074853 Chr.7: 73831555-73831555 on Build GRCh38
rs782476036 Chr.7: 73831726-73831726 on Build GRCh38
rs782664208 Chr.7: 73831663-73831663 on Build GRCh38
rs782776359 Chr.7: 73831730-73831730 on Build GRCh38
rs139694614 Chr.7: 73831699-73831699 on Build GRCh38
rs782399679 Chr.7: 73831682-73831682 on Build GRCh38
rs781852351 Chr.7: 73831652-73831652 on Build GRCh38
rs1049466972 Chr.7: 73831571-73831571 on Build GRCh38
rs782354536 Chr.7: 73831606-73831606 on Build GRCh38
rs557716297 Chr.7: 73831634-73831634 on Build GRCh38
rs782496885 Chr.7: 73831617-73831617 on Build GRCh38
rs570729167 Chr.7: 73831558-73831558 on Build GRCh38
rs201477654 Chr.7: 73831708-73831708 on Build GRCh38
rs781798709 Chr.7: 73831559-73831559 on Build GRCh38
rs782176010 Chr.7: 73831741-73831741 on Build GRCh38
rs371517967 Chr.7: 73831674-73831674 on Build GRCh38
rs782293751 Chr.7: 73831581-73831581 on Build GRCh38
rs782633820 Chr.7: 73831580-73831580 on Build GRCh38
rs781860549 Chr.7: 73831732-73831732 on Build GRCh38
rs782453491 Chr.7: 73831709-73831710 on Build GRCh38
rs148026351 Chr.7: 73831621-73831621 on Build GRCh38
rs782062913 Chr.7: 73831627-73831627 on Build GRCh38
rs376016774 Chr.7: 73831585-73831585 on Build GRCh38
rs1032070026 Chr.7: 73831734-73831734 on Build GRCh38
rs782681015 Chr.7: 73831738-73831738 on Build GRCh38
rs372233301 Chr.7: 73831564-73831564 on Build GRCh38
rs781811608 Chr.7: 73831664-73831664 on Build GRCh38
rs202009827 Chr.7: 73831618-73831618 on Build GRCh38
rs1057401355 Chr.7: 73831668-73831668 on Build GRCh38
rs1037118956 Chr.7: 73831616-73831616 on Build GRCh38
rs968665648 Chr.7: 73831537-73831537 on Build GRCh38
rs575881946 Chr.7: 73831638-73831638 on Build GRCh38
rs367781698 Chr.7: 73831605-73831605 on Build GRCh38
rs143559313 Chr.7: 73831586-73831586 on Build GRCh38
rs200226062 Chr.7: 73831733-73831733 on Build GRCh38
rs375797371 Chr.7: 73831690-73831690 on Build GRCh38
rs138373200 Chr.7: 73831563-73831563 on Build GRCh38
rs575930601 Chr.7: 73831697-73831697 on Build GRCh38
rs781960841 Chr.7: 73831615-73831615 on Build GRCh38
rs782066179 Chr.7: 73831714-73831714 on Build GRCh38
rs922427074 Chr.7: 73831567-73831567 on Build GRCh38
rs782099865 Chr.7: 73831539-73831539 on Build GRCh38
rs146017284 Chr.7: 73831700-73831700 on Build GRCh38
rs782737256 Chr.7: 73831543-73831543 on Build GRCh38
rs782122717 Chr.7: 73831551-73831551 on Build GRCh38
rs781839221 Chr.7: 73831719-73831719 on Build GRCh38
rs781882770 Chr.7: 73831544-73831544 on Build GRCh38
rs782337604 Chr.7: 73831698-73831698 on Build GRCh38
rs909638840 Chr.7: 73831600-73831600 on Build GRCh38
rs782628449 Chr.7: 73831650-73831650 on Build GRCh38
rs782629194 Chr.7: 73831673-73831673 on Build GRCh38
rs782411325 Chr.7: 73831693-73831693 on Build GRCh38
rs34646484 Chr.7: 73831568-73831568 on Build GRCh38

View Full Product Details