Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184276456-184276729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECE2
Entrez Gene ID
9718
Gene Name
endothelin converting enzyme 2
Gene Aliases
-
UniGene
Hs.146161
Gene Chromosome Location
Chr.3: 184249657-184293031 on Build GRCh38
Transcripts
NM_001037324.2 NM_001100120.1 NM_001100121.1 NM_014693.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376446 c.220T>A p.C74S Chr.3: 184276661-184276661 on Build GRCh38
4466737 c.144C>T p.S48S Chr.3: 184276585-184276585 on Build GRCh38
6866934 c.92C>A p.P31H Chr.3: 184276533-184276533 on Build GRCh38
5585079 c.167C>T p.P56L Chr.3: 184276608-184276608 on Build GRCh38
6651285 c.79G>A p.A27T Chr.3: 184276520-184276520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530767382 Chr.3: 184276576-184276576 on Build GRCh38
rs924935279 Chr.3: 184276514-184276514 on Build GRCh38
rs371470060 Chr.3: 184276519-184276519 on Build GRCh38
rs780122559 Chr.3: 184276523-184276523 on Build GRCh38
rs373518539 Chr.3: 184276555-184276555 on Build GRCh38
rs930531858 Chr.3: 184276574-184276574 on Build GRCh38
rs756834004 Chr.3: 184276599-184276599 on Build GRCh38
rs767686541 Chr.3: 184276488-184276488 on Build GRCh38
rs777509624 Chr.3: 184276652-184276652 on Build GRCh38
rs201916543 Chr.3: 184276497-184276497 on Build GRCh38
rs762026206 Chr.3: 184276682-184276682 on Build GRCh38
rs1045477885 Chr.3: 184276622-184276622 on Build GRCh38
rs752393564 Chr.3: 184276645-184276645 on Build GRCh38
rs1030214364 Chr.3: 184276484-184276484 on Build GRCh38
rs747094784 Chr.3: 184276550-184276551 on Build GRCh38
rs912674865 Chr.3: 184276534-184276534 on Build GRCh38
rs777346754 Chr.3: 184276560-184276560 on Build GRCh38
rs913381168 Chr.3: 184276502-184276502 on Build GRCh38
rs764776942 Chr.3: 184276507-184276507 on Build GRCh38
rs775706928 Chr.3: 184276489-184276489 on Build GRCh38
rs906985842 Chr.3: 184276628-184276628 on Build GRCh38
rs997761786 Chr.3: 184276482-184276483 on Build GRCh38
rs766055037 Chr.3: 184276629-184276629 on Build GRCh38
rs376181868 Chr.3: 184276640-184276640 on Build GRCh38
rs781008019 Chr.3: 184276643-184276643 on Build GRCh38
rs770199351 Chr.3: 184276683-184276683 on Build GRCh38
rs369748478 Chr.3: 184276533-184276533 on Build GRCh38
rs768791533 Chr.3: 184276663-184276663 on Build GRCh38
rs751246050 Chr.3: 184276639-184276639 on Build GRCh38
rs757915768 Chr.3: 184276618-184276618 on Build GRCh38
rs773588790 Chr.3: 184276706-184276706 on Build GRCh38
rs145062341 Chr.3: 184276531-184276531 on Build GRCh38
rs13063766 Chr.3: 184276659-184276659 on Build GRCh38
rs746665156 Chr.3: 184276525-184276525 on Build GRCh38
rs1040134230 Chr.3: 184276543-184276543 on Build GRCh38
rs765617903 Chr.3: 184276598-184276598 on Build GRCh38
rs745882263 Chr.3: 184276559-184276559 on Build GRCh38
rs780163260 Chr.3: 184276673-184276673 on Build GRCh38
rs376213717 Chr.3: 184276526-184276526 on Build GRCh38
rs776938670 Chr.3: 184276677-184276677 on Build GRCh38
rs749865348 Chr.3: 184276509-184276509 on Build GRCh38
rs764443817 Chr.3: 184276496-184276496 on Build GRCh38
rs760968421 Chr.3: 184276492-184276492 on Build GRCh38
rs762272689 Chr.3: 184276585-184276585 on Build GRCh38
rs942717079 Chr.3: 184276535-184276535 on Build GRCh38
rs764423218 Chr.3: 184276568-184276568 on Build GRCh38
rs760951995 Chr.3: 184276564-184276564 on Build GRCh38
rs772131814 Chr.3: 184276668-184276668 on Build GRCh38
rs755931516 Chr.3: 184276483-184276483 on Build GRCh38
rs768830091 Chr.3: 184276674-184276674 on Build GRCh38
rs763614116 Chr.3: 184276712-184276712 on Build GRCh38
rs757756789 Chr.3: 184276503-184276503 on Build GRCh38
rs986088545 Chr.3: 184276513-184276513 on Build GRCh38
rs769657755 Chr.3: 184276541-184276541 on Build GRCh38

View Full Product Details