Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49803346-49803591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212A
Entrez Gene ID
389119
Gene Name
family with sequence similarity 212 member A
Gene Aliases
C3orf54, INKA1
UniGene
Hs.86674
Gene Chromosome Location
Chr.3: 49803130-49805030 on Build GRCh38
Transcripts
NM_203370.1 XM_017006372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6010988 c.21G>C p.R7R Chr.3: 49803407-49803407 on Build GRCh38
3824186 c.6C>A p.D2E Chr.3: 49803392-49803392 on Build GRCh38
7192975 c.23T>G p.L8R Chr.3: 49803409-49803409 on Build GRCh38
1246829 c.54_55insCTG p.L20_C21insL Chr.3: 49803440-49803441 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766038420 Chr.3: 49803402-49803402 on Build GRCh38
rs746939856 Chr.3: 49803454-49803454 on Build GRCh38
rs865891089 Chr.3: 49803396-49803396 on Build GRCh38
rs199972862 Chr.3: 49803482-49803482 on Build GRCh38
rs950112237 Chr.3: 49803519-49803519 on Build GRCh38
rs769654497 Chr.3: 49803475-49803475 on Build GRCh38
rs780732547 Chr.3: 49803382-49803382 on Build GRCh38
rs371132855 Chr.3: 49803461-49803461 on Build GRCh38
rs529104886 Chr.3: 49803503-49803503 on Build GRCh38
rs759130418 Chr.3: 49803486-49803486 on Build GRCh38
rs780144524 Chr.3: 49803452-49803452 on Build GRCh38
rs778961736 Chr.3: 49803432-49803432 on Build GRCh38
rs768365016 Chr.3: 49803395-49803395 on Build GRCh38
rs988190844 Chr.3: 49803423-49803423 on Build GRCh38
rs750678309 Chr.3: 49803435-49803435 on Build GRCh38
rs1002119886 Chr.3: 49803386-49803386 on Build GRCh38
rs115355405 Chr.3: 49803410-49803410 on Build GRCh38
rs905067517 Chr.3: 49803372-49803372 on Build GRCh38
rs746754003 Chr.3: 49803375-49803375 on Build GRCh38
rs777224914 Chr.3: 49803493-49803493 on Build GRCh38
rs181384342 Chr.3: 49803553-49803553 on Build GRCh38
rs887624932 Chr.3: 49803544-49803544 on Build GRCh38
rs758447443 Chr.3: 49803450-49803450 on Build GRCh38
rs1044533835 Chr.3: 49803520-49803520 on Build GRCh38
rs895847854 Chr.3: 49803517-49803517 on Build GRCh38
rs748989049 Chr.3: 49803480-49803480 on Build GRCh38
rs1044146948 Chr.3: 49803545-49803545 on Build GRCh38
rs377727330 Chr.3: 49803458-49803458 on Build GRCh38
rs1035715316 Chr.3: 49803394-49803394 on Build GRCh38
rs200735115 Chr.3: 49803479-49803479 on Build GRCh38
rs6807837 Chr.3: 49803490-49803490 on Build GRCh38
rs965088127 Chr.3: 49803428-49803428 on Build GRCh38
rs375706968 Chr.3: 49803470-49803470 on Build GRCh38
rs757643116 Chr.3: 49803414-49803414 on Build GRCh38
rs762322664 Chr.3: 49803397-49803397 on Build GRCh38
rs764540160 Chr.3: 49803408-49803408 on Build GRCh38
rs753224861 Chr.3: 49803407-49803407 on Build GRCh38
rs373219597 Chr.3: 49803368-49803368 on Build GRCh38
rs1020592462 Chr.3: 49803427-49803427 on Build GRCh38
rs777983015 Chr.3: 49803482-49803483 on Build GRCh38

View Full Product Details