Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79032640-79032905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMGDH
Entrez Gene ID
29958
Gene Name
dimethylglycine dehydrogenase
Gene Aliases
DMGDHD, ME2GLYDH
UniGene
Hs.655653
Gene Chromosome Location
Chr.5: 78997564-79069674 on Build GRCh38
Transcripts
NM_013391.3 NR_104002.1 NR_104003.1 XM_006714597.2 XM_011543354.2 XM_011543355.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366170 c.1470G>A p.W490* Chr.5: 79032734-79032734 on Build GRCh38
3856427 c.1500C>T p.G500G Chr.5: 79032704-79032704 on Build GRCh38
3856428 c.897C>T p.G299G Chr.5: 79032704-79032704 on Build GRCh38
450085 c.1510C>T p.Q504* Chr.5: 79032694-79032694 on Build GRCh38
6171942 c.875C>A p.P292Q Chr.5: 79032726-79032726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534488827 Chr.5: 79032803-79032803 on Build GRCh38
rs774921876 Chr.5: 79032809-79032809 on Build GRCh38
rs75001541 Chr.5: 79032859-79032859 on Build GRCh38
rs548607471 Chr.5: 79032759-79032759 on Build GRCh38
rs368017916 Chr.5: 79032795-79032795 on Build GRCh38
rs372669786 Chr.5: 79032804-79032804 on Build GRCh38
rs374196780 Chr.5: 79032663-79032663 on Build GRCh38
rs774773135 Chr.5: 79032730-79032730 on Build GRCh38
rs540045918 Chr.5: 79032875-79032875 on Build GRCh38
rs770670887 Chr.5: 79032683-79032683 on Build GRCh38
rs758123134 Chr.5: 79032850-79032850 on Build GRCh38
rs772825572 Chr.5: 79032799-79032799 on Build GRCh38
rs373980096 Chr.5: 79032771-79032771 on Build GRCh38
rs778707775 Chr.5: 79032687-79032691 on Build GRCh38
rs756348424 Chr.5: 79032744-79032744 on Build GRCh38
rs757059766 Chr.5: 79032786-79032786 on Build GRCh38
rs777884669 Chr.5: 79032670-79032670 on Build GRCh38
rs867784349 Chr.5: 79032849-79032849 on Build GRCh38
rs769394355 Chr.5: 79032797-79032797 on Build GRCh38
rs371277894 Chr.5: 79032802-79032802 on Build GRCh38
rs746100635 Chr.5: 79032694-79032694 on Build GRCh38
rs762287789 Chr.5: 79032819-79032819 on Build GRCh38
rs746770032 Chr.5: 79032674-79032674 on Build GRCh38
rs745676529 Chr.5: 79032796-79032796 on Build GRCh38
rs949520858 Chr.5: 79032722-79032722 on Build GRCh38
rs201191309 Chr.5: 79032855-79032855 on Build GRCh38
rs192307313 Chr.5: 79032775-79032775 on Build GRCh38
rs762211211 Chr.5: 79032735-79032735 on Build GRCh38
rs745609729 Chr.5: 79032743-79032744 on Build GRCh38
rs187836704 Chr.5: 79032687-79032687 on Build GRCh38
rs201067884 Chr.5: 79032874-79032874 on Build GRCh38
rs761155776 Chr.5: 79032823-79032823 on Build GRCh38
rs749394633 Chr.5: 79032660-79032660 on Build GRCh38
rs112410293 Chr.5: 79032704-79032704 on Build GRCh38
rs755104147 Chr.5: 79032848-79032848 on Build GRCh38
rs531571199 Chr.5: 79032752-79032752 on Build GRCh38
rs370857629 Chr.5: 79032764-79032764 on Build GRCh38
rs1055342744 Chr.5: 79032686-79032686 on Build GRCh38
rs769447340 Chr.5: 79032703-79032703 on Build GRCh38
rs757072578 Chr.5: 79032870-79032870 on Build GRCh38
rs267600701 Chr.5: 79032746-79032746 on Build GRCh38
rs762477431 Chr.5: 79032725-79032725 on Build GRCh38
rs750647748 Chr.5: 79032743-79032743 on Build GRCh38
rs1805072 Chr.5: 79032737-79032737 on Build GRCh38
rs768086517 Chr.5: 79032820-79032820 on Build GRCh38
rs766660104 Chr.5: 79032842-79032842 on Build GRCh38
rs1038625096 Chr.5: 79032668-79032668 on Build GRCh38
rs774911260 Chr.5: 79032724-79032724 on Build GRCh38
rs148916205 Chr.5: 79032726-79032726 on Build GRCh38
rs1020148307 Chr.5: 79032841-79032841 on Build GRCh38

View Full Product Details