Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154449234-154449481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM50A
Entrez Gene ID
9130
Gene Name
family with sequence similarity 50 member A
Gene Aliases
9F, DXS9928E, HXC-26, HXC26, XAP5
UniGene
Hs.54277
Gene Chromosome Location
Chr.X: 154444126-154450654 on Build GRCh38
Transcripts
NM_004699.3

SNP Detail

SNP ID Chromosome Location
rs989621282 Chr.X: 154449306-154449306 on Build GRCh38
rs782370131 Chr.X: 154449361-154449361 on Build GRCh38
rs782443455 Chr.X: 154449320-154449320 on Build GRCh38
rs782504746 Chr.X: 154449333-154449333 on Build GRCh38
rs149398479 Chr.X: 154449381-154449381 on Build GRCh38
rs373011856 Chr.X: 154449338-154449338 on Build GRCh38
rs782485112 Chr.X: 154449292-154449292 on Build GRCh38
rs900236826 Chr.X: 154449421-154449421 on Build GRCh38
rs782582096 Chr.X: 154449311-154449312 on Build GRCh38
rs781869434 Chr.X: 154449310-154449310 on Build GRCh38
rs1029977623 Chr.X: 154449439-154449439 on Build GRCh38
rs376772251 Chr.X: 154449309-154449309 on Build GRCh38
rs2070643 Chr.X: 154449352-154449352 on Build GRCh38
rs369805936 Chr.X: 154449328-154449328 on Build GRCh38
rs782419846 Chr.X: 154449340-154449340 on Build GRCh38
rs782453073 Chr.X: 154449293-154449294 on Build GRCh38
rs782268721 Chr.X: 154449403-154449403 on Build GRCh38
rs888648505 Chr.X: 154449398-154449398 on Build GRCh38
rs782277025 Chr.X: 154449313-154449313 on Build GRCh38
rs781907318 Chr.X: 154449325-154449325 on Build GRCh38
rs782718618 Chr.X: 154449275-154449275 on Build GRCh38
rs941521430 Chr.X: 154449418-154449418 on Build GRCh38
rs782127107 Chr.X: 154449455-154449455 on Build GRCh38
rs1037479885 Chr.X: 154449420-154449420 on Build GRCh38
rs930190095 Chr.X: 154449375-154449375 on Build GRCh38
rs781788063 Chr.X: 154449288-154449288 on Build GRCh38
rs782392847 Chr.X: 154449330-154449330 on Build GRCh38
rs782651569 Chr.X: 154449350-154449350 on Build GRCh38
rs144474497 Chr.X: 154449265-154449265 on Build GRCh38
rs781975942 Chr.X: 154449448-154449448 on Build GRCh38
rs115202723 Chr.X: 154449430-154449430 on Build GRCh38

View Full Product Details