Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122275947-122276182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
UniGene
Hs.435615
Gene Chromosome Location
Chr.3: 122183683-122286503 on Build GRCh38
Transcripts
NM_000388.3 NM_001178065.1 XM_005247837.2 XM_006713789.3 XM_017007324.1 XM_017007325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620676 c.1571A>T p.N524I Chr.3: 122276005-122276005 on Build GRCh38
3695684 c.1604G>T p.R535M Chr.3: 122276038-122276038 on Build GRCh38
6891121 c.1519G>A p.E507K Chr.3: 122275953-122275953 on Build GRCh38
5799294 c.1604G>A p.R535K Chr.3: 122276038-122276038 on Build GRCh38
3586253 c.1553G>A p.G518E Chr.3: 122275987-122275987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776904675 Chr.3: 122275994-122275994 on Build GRCh38
rs2279802 Chr.3: 122276094-122276094 on Build GRCh38
rs201568219 Chr.3: 122276010-122276010 on Build GRCh38
rs767994162 Chr.3: 122276053-122276053 on Build GRCh38
rs753432681 Chr.3: 122276080-122276080 on Build GRCh38
rs199688157 Chr.3: 122276007-122276007 on Build GRCh38
rs746054548 Chr.3: 122275998-122275998 on Build GRCh38
rs764669682 Chr.3: 122276026-122276026 on Build GRCh38
rs878853974 Chr.3: 122276046-122276046 on Build GRCh38
rs780400116 Chr.3: 122276058-122276059 on Build GRCh38
rs768860003 Chr.3: 122276066-122276067 on Build GRCh38
rs768663095 Chr.3: 122275983-122275983 on Build GRCh38
rs778966330 Chr.3: 122276092-122276092 on Build GRCh38
rs753082320 Chr.3: 122276131-122276131 on Build GRCh38
rs562956195 Chr.3: 122276141-122276141 on Build GRCh38
rs138099808 Chr.3: 122275988-122275988 on Build GRCh38
rs762299355 Chr.3: 122276039-122276039 on Build GRCh38
rs750903679 Chr.3: 122276049-122276050 on Build GRCh38
rs565299549 Chr.3: 122276054-122276054 on Build GRCh38
rs762899043 Chr.3: 122276021-122276021 on Build GRCh38
rs898193456 Chr.3: 122276009-122276009 on Build GRCh38
rs1034006357 Chr.3: 122276145-122276146 on Build GRCh38
rs548601727 Chr.3: 122276153-122276153 on Build GRCh38
rs779081482 Chr.3: 122276047-122276048 on Build GRCh38
rs559849402 Chr.3: 122276071-122276071 on Build GRCh38
rs377233360 Chr.3: 122275987-122275987 on Build GRCh38
rs754387056 Chr.3: 122276084-122276084 on Build GRCh38
rs531889875 Chr.3: 122276142-122276142 on Build GRCh38
rs757438825 Chr.3: 122276046-122276047 on Build GRCh38
rs778647030 Chr.3: 122275985-122275985 on Build GRCh38
rs993925299 Chr.3: 122275991-122275991 on Build GRCh38
rs527648150 Chr.3: 122276080-122276080 on Build GRCh38
rs532054744 Chr.3: 122276078-122276078 on Build GRCh38
rs200470956 Chr.3: 122276008-122276008 on Build GRCh38
rs769907714 Chr.3: 122276000-122276000 on Build GRCh38
rs750141663 Chr.3: 122276069-122276069 on Build GRCh38
rs765606865 Chr.3: 122276149-122276149 on Build GRCh38
rs774666667 Chr.3: 122276030-122276030 on Build GRCh38
rs755073303 Chr.3: 122276044-122276044 on Build GRCh38
rs747438512 Chr.3: 122276061-122276064 on Build GRCh38
rs758998412 Chr.3: 122276056-122276057 on Build GRCh38
rs1028068213 Chr.3: 122276067-122276067 on Build GRCh38
rs776534485 Chr.3: 122276006-122276006 on Build GRCh38

View Full Product Details