Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73192503-73192734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149B1
Entrez Gene ID
317662
Gene Name
family with sequence similarity 149 member B1
Gene Aliases
KIAA0974
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73168090-73242181 on Build GRCh38
Transcripts
NM_173348.1 XM_005269744.2 XM_005269745.2 XM_005269747.3 XM_011539737.2 XM_011539740.2 XM_017016164.1 XM_017016165.1 XM_017016166.1 XM_017016167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3709897 c.321G>T p.M107I Chr.10: 73192594-73192594 on Build GRCh38
4135660 c.313C>A p.Q105K Chr.10: 73192586-73192586 on Build GRCh38
7367075 c.349G>C p.E117Q Chr.10: 73192622-73192622 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757063365 Chr.10: 73192616-73192616 on Build GRCh38
rs1024747746 Chr.10: 73192645-73192645 on Build GRCh38
rs763616242 Chr.10: 73192538-73192538 on Build GRCh38
rs1009666232 Chr.10: 73192532-73192532 on Build GRCh38
rs892314739 Chr.10: 73192588-73192588 on Build GRCh38
rs1009293598 Chr.10: 73192604-73192604 on Build GRCh38
rs967206111 Chr.10: 73192618-73192618 on Build GRCh38
rs187496569 Chr.10: 73192639-73192639 on Build GRCh38
rs760398523 Chr.10: 73192537-73192537 on Build GRCh38
rs750456652 Chr.10: 73192710-73192710 on Build GRCh38
rs753693466 Chr.10: 73192591-73192591 on Build GRCh38
rs559604779 Chr.10: 73192540-73192540 on Build GRCh38
rs1032963957 Chr.10: 73192649-73192649 on Build GRCh38

View Full Product Details