Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108510855-108511122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
UniGene
Hs.28540
Gene Chromosome Location
Chr.11: 108505431-108607514 on Build GRCh38
Transcripts
NM_001144763.1 NM_001144764.1 NM_001144765.1 NM_001308019.1 NM_015065.2 XM_011542696.2 XM_011542698.2 XM_011542700.2 XM_017017397.1 XM_017017398.1 XM_017017399.1 XM_017017400.1 XM_017017401.1 XM_017017402.1 XM_017017403.1 XM_017017404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4145213 c.4452C>A p.G1484G Chr.11: 108511055-108511055 on Build GRCh38
5429733 c.4597C>G p.L1533V Chr.11: 108510910-108510910 on Build GRCh38
3752122 c.4590A>G p.L1530L Chr.11: 108510917-108510917 on Build GRCh38
6657330 c.4419T>C p.G1473G Chr.11: 108511088-108511088 on Build GRCh38
2110881 c.4454G>A p.R1485K Chr.11: 108511053-108511053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764925317 Chr.11: 108511001-108511001 on Build GRCh38
rs768150921 Chr.11: 108511066-108511066 on Build GRCh38
rs765850451 Chr.11: 108510916-108510917 on Build GRCh38
rs200270555 Chr.11: 108510892-108510892 on Build GRCh38
rs761804692 Chr.11: 108510993-108510993 on Build GRCh38
rs759792392 Chr.11: 108511080-108511080 on Build GRCh38
rs751045536 Chr.11: 108510998-108511001 on Build GRCh38
rs983720236 Chr.11: 108510932-108510932 on Build GRCh38
rs777965883 Chr.11: 108511034-108511034 on Build GRCh38
rs750273457 Chr.11: 108511006-108511006 on Build GRCh38
rs760124476 Chr.11: 108510982-108510982 on Build GRCh38
rs139262980 Chr.11: 108510904-108510904 on Build GRCh38
rs906546463 Chr.11: 108511025-108511025 on Build GRCh38
rs955760001 Chr.11: 108511050-108511050 on Build GRCh38
rs149702505 Chr.11: 108510988-108510988 on Build GRCh38
rs571085648 Chr.11: 108510910-108510910 on Build GRCh38
rs747047365 Chr.11: 108510939-108510939 on Build GRCh38
rs376594541 Chr.11: 108510882-108510882 on Build GRCh38
rs916651272 Chr.11: 108510952-108510952 on Build GRCh38
rs775034564 Chr.11: 108510973-108510973 on Build GRCh38
rs150950878 Chr.11: 108510943-108510943 on Build GRCh38
rs777124686 Chr.11: 108510980-108510980 on Build GRCh38
rs765181920 Chr.11: 108511093-108511093 on Build GRCh38
rs776174830 Chr.11: 108511073-108511073 on Build GRCh38
rs753792351 Chr.11: 108510888-108510888 on Build GRCh38
rs989838172 Chr.11: 108511090-108511090 on Build GRCh38
rs760324571 Chr.11: 108511056-108511056 on Build GRCh38
rs1030005399 Chr.11: 108510881-108510881 on Build GRCh38
rs79942405 Chr.11: 108510917-108510917 on Build GRCh38
rs763603409 Chr.11: 108510987-108510987 on Build GRCh38
rs368977137 Chr.11: 108510945-108510945 on Build GRCh38
rs748672486 Chr.11: 108510949-108510949 on Build GRCh38
rs140766467 Chr.11: 108510951-108510951 on Build GRCh38
rs780274167 Chr.11: 108511008-108511008 on Build GRCh38
rs778774056 Chr.11: 108510906-108510906 on Build GRCh38
rs565800352 Chr.11: 108510916-108510916 on Build GRCh38
rs749623073 Chr.11: 108511042-108511042 on Build GRCh38
rs755045088 Chr.11: 108511011-108511011 on Build GRCh38
rs371382968 Chr.11: 108511019-108511019 on Build GRCh38
rs773687109 Chr.11: 108510954-108510954 on Build GRCh38
rs767235950 Chr.11: 108510968-108510968 on Build GRCh38
rs759091170 Chr.11: 108510956-108510956 on Build GRCh38
rs771218637 Chr.11: 108511047-108511047 on Build GRCh38
rs763975244 Chr.11: 108510883-108510883 on Build GRCh38
rs781493293 Chr.11: 108511015-108511015 on Build GRCh38
rs574611078 Chr.11: 108511016-108511016 on Build GRCh38
rs758161398 Chr.11: 108511007-108511007 on Build GRCh38
rs775062364 Chr.11: 108511049-108511049 on Build GRCh38
rs1046796800 Chr.11: 108511000-108511000 on Build GRCh38

View Full Product Details