Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63764472-63764745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf95
Entrez Gene ID
65998
Gene Name
chromosome 11 open reading frame 95
Gene Aliases
-
UniGene
Hs.191073
Gene Chromosome Location
Chr.11: 63759892-63768641 on Build GRCh38
Transcripts
NM_001144936.1

SNP Detail

SNP ID Chromosome Location
rs934163883 Chr.11: 63764581-63764583 on Build GRCh38
rs1002108664 Chr.11: 63764726-63764726 on Build GRCh38
rs556757135 Chr.11: 63764705-63764705 on Build GRCh38
rs755480784 Chr.11: 63764545-63764545 on Build GRCh38
rs910252886 Chr.11: 63764508-63764508 on Build GRCh38
rs893908623 Chr.11: 63764591-63764591 on Build GRCh38
rs942536649 Chr.11: 63764559-63764559 on Build GRCh38
rs941489135 Chr.11: 63764664-63764664 on Build GRCh38
rs570282619 Chr.11: 63764708-63764708 on Build GRCh38
rs1047109729 Chr.11: 63764586-63764586 on Build GRCh38
rs887197319 Chr.11: 63764625-63764625 on Build GRCh38
rs536758056 Chr.11: 63764643-63764643 on Build GRCh38
rs547813253 Chr.11: 63764506-63764506 on Build GRCh38
rs911018248 Chr.11: 63764557-63764557 on Build GRCh38
rs567604366 Chr.11: 63764628-63764628 on Build GRCh38
rs972412622 Chr.11: 63764510-63764510 on Build GRCh38
rs1039073919 Chr.11: 63764584-63764584 on Build GRCh38
rs918269689 Chr.11: 63764513-63764513 on Build GRCh38
rs1003791313 Chr.11: 63764714-63764714 on Build GRCh38
rs902552929 Chr.11: 63764703-63764703 on Build GRCh38
rs962662977 Chr.11: 63764503-63764503 on Build GRCh38
rs1050995667 Chr.11: 63764549-63764549 on Build GRCh38
rs942915887 Chr.11: 63764518-63764518 on Build GRCh38

View Full Product Details