Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33025862-33026091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC7
Entrez Gene ID
91614
Gene Name
DEP domain containing 7
Gene Aliases
TR2, dJ85M6.4
UniGene
Hs.280990
Gene Chromosome Location
Chr.11: 33015864-33033582 on Build GRCh38
Transcripts
NM_001077242.1 NM_139160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2114806 c.271C>G p.Q91E Chr.11: 33025883-33025883 on Build GRCh38
4889573 c.365C>G p.P122R Chr.11: 33025950-33025950 on Build GRCh38
2114808 c.275C>T p.A92V Chr.11: 33025887-33025887 on Build GRCh38
2114816 c.437+2T>C p.? Chr.11: 33026051-33026051 on Build GRCh38
6537296 c.355A>G p.S119G Chr.11: 33025967-33025967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753120869 Chr.11: 33025899-33025899 on Build GRCh38
rs781700415 Chr.11: 33025949-33025949 on Build GRCh38
rs754187737 Chr.11: 33025940-33025940 on Build GRCh38
rs991189900 Chr.11: 33026024-33026024 on Build GRCh38
rs773955423 Chr.11: 33026018-33026018 on Build GRCh38
rs749467283 Chr.11: 33026013-33026016 on Build GRCh38
rs756567724 Chr.11: 33026008-33026019 on Build GRCh38
rs780285915 Chr.11: 33026008-33026011 on Build GRCh38
rs781243212 Chr.11: 33025968-33025968 on Build GRCh38
rs200889394 Chr.11: 33026065-33026065 on Build GRCh38
rs374632275 Chr.11: 33026036-33026036 on Build GRCh38
rs750559110 Chr.11: 33025960-33025960 on Build GRCh38
rs142562030 Chr.11: 33026058-33026058 on Build GRCh38
rs761157222 Chr.11: 33025916-33025916 on Build GRCh38
rs866171135 Chr.11: 33025989-33025989 on Build GRCh38
rs888419252 Chr.11: 33025934-33025934 on Build GRCh38
rs764361404 Chr.11: 33025931-33025931 on Build GRCh38
rs372513278 Chr.11: 33025887-33025887 on Build GRCh38
rs764600089 Chr.11: 33026062-33026062 on Build GRCh38
rs531309178 Chr.11: 33026045-33026045 on Build GRCh38
rs754273487 Chr.11: 33026063-33026063 on Build GRCh38
rs977086642 Chr.11: 33025948-33025948 on Build GRCh38
rs376814687 Chr.11: 33025956-33025956 on Build GRCh38
rs758998296 Chr.11: 33025888-33025888 on Build GRCh38
rs576295157 Chr.11: 33026032-33026032 on Build GRCh38
rs750740480 Chr.11: 33025997-33025997 on Build GRCh38
rs573702016 Chr.11: 33025944-33025944 on Build GRCh38
rs556333291 Chr.11: 33025978-33025978 on Build GRCh38
rs771872138 Chr.11: 33025972-33025972 on Build GRCh38
rs770594618 Chr.11: 33026017-33026017 on Build GRCh38
rs202173561 Chr.11: 33025975-33025975 on Build GRCh38
rs749025119 Chr.11: 33025984-33025984 on Build GRCh38
rs371496330 Chr.11: 33025965-33025965 on Build GRCh38
rs757731519 Chr.11: 33025942-33025943 on Build GRCh38
rs771468155 Chr.11: 33026042-33026042 on Build GRCh38
rs768102857 Chr.11: 33025889-33025889 on Build GRCh38
rs748025899 Chr.11: 33025974-33025974 on Build GRCh38
rs377735139 Chr.11: 33026052-33026052 on Build GRCh38

View Full Product Details