Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 77801338-77801569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIGA1
Entrez Gene ID
374986
Gene Name
mitoguardin 1
Gene Aliases
FAM73A
UniGene
Hs.437755
Gene Chromosome Location
Chr.1: 77779624-77879540 on Build GRCh38
Transcripts
NM_001270384.1 NM_198549.3 XM_005270846.4 XM_005270847.4 XM_005270848.4 XM_011541422.2 XM_017001227.1 XM_017001228.1 XM_017001229.1 XM_017001230.1 XM_017001231.1 XM_017001232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
912295 c.284G>T p.R95I Chr.1: 77801419-77801419 on Build GRCh38
1296711 c.318A>G p.P106P Chr.1: 77801453-77801453 on Build GRCh38
1344404 c.290G>A p.R97H Chr.1: 77801425-77801425 on Build GRCh38
5381556 c.211G>A p.V71M Chr.1: 77801346-77801346 on Build GRCh38
5587778 c.331C>A p.H111N Chr.1: 77801466-77801466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367766355 Chr.1: 77801465-77801465 on Build GRCh38
rs145668210 Chr.1: 77801484-77801484 on Build GRCh38
rs746573349 Chr.1: 77801371-77801371 on Build GRCh38
rs752541579 Chr.1: 77801464-77801464 on Build GRCh38
rs530750133 Chr.1: 77801452-77801452 on Build GRCh38
rs746376147 Chr.1: 77801377-77801377 on Build GRCh38
rs766210952 Chr.1: 77801425-77801425 on Build GRCh38
rs752682706 Chr.1: 77801528-77801529 on Build GRCh38
rs772118680 Chr.1: 77801532-77801532 on Build GRCh38
rs774540541 Chr.1: 77801410-77801410 on Build GRCh38
rs751110689 Chr.1: 77801518-77801519 on Build GRCh38
rs569952814 Chr.1: 77801375-77801375 on Build GRCh38
rs764966876 Chr.1: 77801462-77801462 on Build GRCh38
rs767205893 Chr.1: 77801526-77801529 on Build GRCh38
rs749708562 Chr.1: 77801512-77801512 on Build GRCh38
rs374197863 Chr.1: 77801388-77801388 on Build GRCh38
rs140911803 Chr.1: 77801478-77801478 on Build GRCh38
rs781753466 Chr.1: 77801376-77801376 on Build GRCh38
rs114177761 Chr.1: 77801472-77801472 on Build GRCh38
rs776027314 Chr.1: 77801387-77801387 on Build GRCh38
rs535909619 Chr.1: 77801508-77801508 on Build GRCh38
rs374487616 Chr.1: 77801522-77801522 on Build GRCh38
rs746346780 Chr.1: 77801493-77801493 on Build GRCh38
rs770445172 Chr.1: 77801502-77801502 on Build GRCh38
rs762017362 Chr.1: 77801413-77801413 on Build GRCh38
rs758357008 Chr.1: 77801530-77801530 on Build GRCh38
rs942268713 Chr.1: 77801368-77801368 on Build GRCh38
rs374261295 Chr.1: 77801394-77801394 on Build GRCh38
rs759392225 Chr.1: 77801453-77801453 on Build GRCh38
rs900690777 Chr.1: 77801384-77801384 on Build GRCh38
rs777245811 Chr.1: 77801364-77801364 on Build GRCh38
rs758017441 Chr.1: 77801468-77801468 on Build GRCh38
rs763218684 Chr.1: 77801393-77801393 on Build GRCh38
rs1056969968 Chr.1: 77801492-77801492 on Build GRCh38
rs1021710315 Chr.1: 77801518-77801518 on Build GRCh38
rs754872741 Chr.1: 77801520-77801521 on Build GRCh38
rs751020505 Chr.1: 77801476-77801476 on Build GRCh38
rs774769427 Chr.1: 77801528-77801528 on Build GRCh38
rs770129855 Chr.1: 77801385-77801385 on Build GRCh38

View Full Product Details