Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75385825-75386098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3BP
Entrez Gene ID
10087
Gene Name
collagen type IV alpha 3 binding protein
Gene Aliases
CERT, CERTL, GPBP, MRD34, STARD11
UniGene
Hs.270437
Gene Chromosome Location
Chr.5: 75368486-75511981 on Build GRCh38
Transcripts
NM_001130105.1 NM_005713.2 NM_031361.2 XM_006714513.2 XM_011543090.2 XM_011543091.2 XM_017008919.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027088 c.1787G>A p.R596H Chr.5: 75385916-75385916 on Build GRCh38
4621196 c.1786C>T p.R596C Chr.5: 75385917-75385917 on Build GRCh38
196155 c.1398C>T p.D466D Chr.5: 75385921-75385921 on Build GRCh38
3856298 c.1782C>T p.D594D Chr.5: 75385921-75385921 on Build GRCh38
6633240 c.1407T>A p.N469K Chr.5: 75385912-75385912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746481257 Chr.5: 75385925-75385925 on Build GRCh38
rs370396757 Chr.5: 75386052-75386052 on Build GRCh38
rs375406284 Chr.5: 75386018-75386018 on Build GRCh38
rs757647793 Chr.5: 75386059-75386062 on Build GRCh38
rs149889328 Chr.5: 75385914-75385914 on Build GRCh38
rs770590298 Chr.5: 75385926-75385926 on Build GRCh38
rs370676044 Chr.5: 75385920-75385920 on Build GRCh38
rs757961292 Chr.5: 75385918-75385918 on Build GRCh38
rs763424815 Chr.5: 75385999-75385999 on Build GRCh38
rs770401308 Chr.5: 75385963-75385963 on Build GRCh38
rs61754509 Chr.5: 75386019-75386019 on Build GRCh38
rs769792172 Chr.5: 75385866-75385866 on Build GRCh38
rs764260078 Chr.5: 75386002-75386002 on Build GRCh38
rs144229842 Chr.5: 75386037-75386037 on Build GRCh38
rs748030804 Chr.5: 75385917-75385917 on Build GRCh38
rs761822745 Chr.5: 75386026-75386026 on Build GRCh38
rs698912 Chr.5: 75385948-75385948 on Build GRCh38
rs775705710 Chr.5: 75385996-75385996 on Build GRCh38
rs148645687 Chr.5: 75386032-75386032 on Build GRCh38
rs777222556 Chr.5: 75385936-75385936 on Build GRCh38
rs765265419 Chr.5: 75386062-75386062 on Build GRCh38
rs1003399612 Chr.5: 75386065-75386065 on Build GRCh38
rs34213666 Chr.5: 75385875-75385876 on Build GRCh38
rs1012873977 Chr.5: 75385973-75385973 on Build GRCh38
rs754733713 Chr.5: 75385899-75385899 on Build GRCh38
rs974368321 Chr.5: 75386008-75386008 on Build GRCh38

View Full Product Details