Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151520093-151520360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGN
Entrez Gene ID
57530
Gene Name
cingulin
Gene Aliases
-
UniGene
Hs.591464
Gene Chromosome Location
Chr.1: 151510859-151538692 on Build GRCh38
Transcripts
NM_020770.2 XM_005245365.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3667345 c.956A>G p.Y319C Chr.1: 151520248-151520248 on Build GRCh38
1688533 c.919C>T p.P307S Chr.1: 151520211-151520211 on Build GRCh38
6405307 c.881C>T p.S294L Chr.1: 151520173-151520173 on Build GRCh38
1472607 c.964C>T p.L322L Chr.1: 151520256-151520256 on Build GRCh38
5547869 c.974+7delG p.? Chr.1: 151520273-151520273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759464490 Chr.1: 151520295-151520295 on Build GRCh38
rs766712140 Chr.1: 151520118-151520119 on Build GRCh38
rs756844992 Chr.1: 151520135-151520138 on Build GRCh38
rs899769011 Chr.1: 151520212-151520212 on Build GRCh38
rs929766325 Chr.1: 151520134-151520134 on Build GRCh38
rs768745636 Chr.1: 151520145-151520145 on Build GRCh38
rs763872124 Chr.1: 151520135-151520135 on Build GRCh38
rs762624652 Chr.1: 151520151-151520151 on Build GRCh38
rs984156304 Chr.1: 151520184-151520184 on Build GRCh38
rs750786475 Chr.1: 151520265-151520265 on Build GRCh38
rs1001412032 Chr.1: 151520131-151520141 on Build GRCh38
rs768633353 Chr.1: 151520306-151520308 on Build GRCh38
rs753352882 Chr.1: 151520134-151520135 on Build GRCh38
rs376534912 Chr.1: 151520270-151520270 on Build GRCh38
rs757581587 Chr.1: 151520125-151520125 on Build GRCh38
rs755142038 Chr.1: 151520278-151520278 on Build GRCh38
rs748351952 Chr.1: 151520281-151520281 on Build GRCh38
rs149076431 Chr.1: 151520224-151520224 on Build GRCh38
rs35469373 Chr.1: 151520277-151520278 on Build GRCh38
rs776961078 Chr.1: 151520147-151520147 on Build GRCh38
rs529199632 Chr.1: 151520119-151520122 on Build GRCh38
rs765159175 Chr.1: 151520137-151520138 on Build GRCh38
rs892303538 Chr.1: 151520264-151520264 on Build GRCh38
rs559760884 Chr.1: 151520277-151520277 on Build GRCh38
rs770808876 Chr.1: 151520154-151520154 on Build GRCh38
rs779750347 Chr.1: 151520151-151520152 on Build GRCh38
rs746332873 Chr.1: 151520190-151520190 on Build GRCh38
rs770146427 Chr.1: 151520287-151520287 on Build GRCh38
rs79579671 Chr.1: 151520165-151520165 on Build GRCh38
rs200681495 Chr.1: 151520170-151520170 on Build GRCh38
rs199587739 Chr.1: 151520251-151520251 on Build GRCh38
rs202137727 Chr.1: 151520237-151520237 on Build GRCh38
rs375594870 Chr.1: 151520135-151520135 on Build GRCh38
rs747176969 Chr.1: 151520139-151520139 on Build GRCh38
rs72997944 Chr.1: 151520138-151520138 on Build GRCh38
rs138026419 Chr.1: 151520198-151520198 on Build GRCh38
rs753493702 Chr.1: 151520234-151520234 on Build GRCh38
rs758899903 Chr.1: 151520266-151520266 on Build GRCh38
rs758186584 Chr.1: 151520152-151520152 on Build GRCh38
rs767631841 Chr.1: 151520205-151520205 on Build GRCh38
rs369040309 Chr.1: 151520314-151520314 on Build GRCh38
rs368938931 Chr.1: 151520291-151520291 on Build GRCh38
rs768818595 Chr.1: 151520279-151520279 on Build GRCh38
rs905741352 Chr.1: 151520128-151520128 on Build GRCh38
rs759480796 Chr.1: 151520191-151520191 on Build GRCh38
rs760446063 Chr.1: 151520131-151520147 on Build GRCh38
rs760284344 Chr.1: 151520226-151520226 on Build GRCh38
rs34671806 Chr.1: 151520256-151520256 on Build GRCh38
rs772114789 Chr.1: 151520308-151520308 on Build GRCh38
rs373396830 Chr.1: 151520280-151520280 on Build GRCh38
rs750381907 Chr.1: 151520140-151520145 on Build GRCh38

View Full Product Details