Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226596665-226596924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STUM
Entrez Gene ID
375057
Gene Name
stum, mechanosensory transduction mediator homolog
Gene Aliases
C1orf95
UniGene
Hs.116827
Gene Chromosome Location
Chr.1: 226548238-226609214 on Build GRCh38
Transcripts
NM_001003665.3 XM_011544183.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5352885 c.293C>T p.A98V Chr.1: 226596892-226596892 on Build GRCh38
3671639 c.203-1G>T p.? Chr.1: 226596801-226596801 on Build GRCh38
69927 c.293C>A p.A98E Chr.1: 226596892-226596892 on Build GRCh38
6615363 c.295G>T p.A99S Chr.1: 226596894-226596894 on Build GRCh38
3484282 c.324C>T p.I108I Chr.1: 226596923-226596923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572468782 Chr.1: 226596683-226596683 on Build GRCh38
rs753547083 Chr.1: 226596839-226596839 on Build GRCh38
rs746342425 Chr.1: 226596853-226596853 on Build GRCh38
rs763091873 Chr.1: 226596809-226596809 on Build GRCh38
rs966038399 Chr.1: 226596829-226596829 on Build GRCh38
rs563902383 Chr.1: 226596900-226596900 on Build GRCh38
rs146333541 Chr.1: 226596851-226596851 on Build GRCh38
rs924285158 Chr.1: 226596888-226596888 on Build GRCh38
rs761880064 Chr.1: 226596823-226596823 on Build GRCh38
rs561555576 Chr.1: 226596761-226596761 on Build GRCh38
rs766158260 Chr.1: 226596836-226596836 on Build GRCh38
rs778701361 Chr.1: 226596841-226596841 on Build GRCh38
rs767398022 Chr.1: 226596825-226596825 on Build GRCh38
rs752294585 Chr.1: 226596845-226596845 on Build GRCh38
rs749638369 Chr.1: 226596856-226596856 on Build GRCh38
rs771630623 Chr.1: 226596764-226596764 on Build GRCh38
rs746266197 Chr.1: 226596768-226596768 on Build GRCh38
rs770437978 Chr.1: 226596765-226596765 on Build GRCh38
rs188943559 Chr.1: 226596785-226596785 on Build GRCh38
rs1010988376 Chr.1: 226596689-226596689 on Build GRCh38
rs774473845 Chr.1: 226596815-226596815 on Build GRCh38
rs981189892 Chr.1: 226596875-226596875 on Build GRCh38
rs774526817 Chr.1: 226596883-226596883 on Build GRCh38
rs775601645 Chr.1: 226596800-226596800 on Build GRCh38
rs995421107 Chr.1: 226596733-226596733 on Build GRCh38
rs776251071 Chr.1: 226596810-226596810 on Build GRCh38
rs3738714 Chr.1: 226596685-226596685 on Build GRCh38
rs111228990 Chr.1: 226596833-226596833 on Build GRCh38
rs761931098 Chr.1: 226596884-226596884 on Build GRCh38
rs886543406 Chr.1: 226596752-226596752 on Build GRCh38
rs139902713 Chr.1: 226596873-226596873 on Build GRCh38
rs374465963 Chr.1: 226596846-226596846 on Build GRCh38
rs770146224 Chr.1: 226596774-226596774 on Build GRCh38
rs374590755 Chr.1: 226596758-226596758 on Build GRCh38
rs377114902 Chr.1: 226596904-226596904 on Build GRCh38
rs923775202 Chr.1: 226596863-226596863 on Build GRCh38
rs201231970 Chr.1: 226596854-226596854 on Build GRCh38
rs754621069 Chr.1: 226596840-226596840 on Build GRCh38
rs1039801570 Chr.1: 226596739-226596739 on Build GRCh38
rs760409843 Chr.1: 226596834-226596834 on Build GRCh38

View Full Product Details