Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141516692-141516965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH1
Entrez Gene ID
1729
Gene Name
diaphanous related formin 1
Gene Aliases
DFNA1, DIA1, DRF1, LFHL1, SCBMS, hDIA1
UniGene
Hs.529451
Gene Chromosome Location
Chr.5: 141515021-141619055 on Build GRCh38
Transcripts
NM_001079812.2 NM_001314007.1 NM_005219.4 XM_011537572.2 XM_011537573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5778099 c.3817T>C p.*1273Q Chr.5: 141516853-141516853 on Build GRCh38
5763518 c.3808C>T p.R1270C Chr.5: 141516862-141516862 on Build GRCh38
7285311 c.3809delG p.R1270fs*>3 Chr.5: 141516861-141516861 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368263785 Chr.5: 141516846-141516846 on Build GRCh38
rs762729981 Chr.5: 141516785-141516785 on Build GRCh38
rs774815702 Chr.5: 141516939-141516939 on Build GRCh38
rs761332072 Chr.5: 141516856-141516856 on Build GRCh38
rs199867769 Chr.5: 141516843-141516843 on Build GRCh38
rs371664456 Chr.5: 141516861-141516861 on Build GRCh38
rs753530153 Chr.5: 141516784-141516784 on Build GRCh38
rs926540554 Chr.5: 141516754-141516754 on Build GRCh38
rs753432442 Chr.5: 141516852-141516852 on Build GRCh38
rs749871871 Chr.5: 141516934-141516934 on Build GRCh38
rs758272814 Chr.5: 141516862-141516862 on Build GRCh38
rs770002473 Chr.5: 141516927-141516927 on Build GRCh38
rs747267088 Chr.5: 141516822-141516822 on Build GRCh38
rs777943998 Chr.5: 141516930-141516930 on Build GRCh38
rs554410556 Chr.5: 141516725-141516727 on Build GRCh38
rs773890622 Chr.5: 141516837-141516837 on Build GRCh38
rs751282493 Chr.5: 141516898-141516898 on Build GRCh38
rs554541659 Chr.5: 141516723-141516723 on Build GRCh38
rs1031662096 Chr.5: 141516868-141516868 on Build GRCh38
rs368620770 Chr.5: 141516813-141516813 on Build GRCh38
rs962514448 Chr.5: 141516738-141516738 on Build GRCh38
rs546064158 Chr.5: 141516903-141516903 on Build GRCh38
rs574596779 Chr.5: 141516763-141516763 on Build GRCh38
rs372638311 Chr.5: 141516819-141516819 on Build GRCh38
rs748341361 Chr.5: 141516912-141516912 on Build GRCh38
rs202086273 Chr.5: 141516835-141516835 on Build GRCh38
rs35755269 Chr.5: 141516905-141516905 on Build GRCh38
rs763491790 Chr.5: 141516842-141516842 on Build GRCh38
rs374910187 Chr.5: 141516897-141516897 on Build GRCh38
rs577050340 Chr.5: 141516849-141516849 on Build GRCh38
rs768757611 Chr.5: 141516829-141516829 on Build GRCh38
rs781375115 Chr.5: 141516834-141516834 on Build GRCh38
rs757222276 Chr.5: 141516818-141516818 on Build GRCh38
rs771335028 Chr.5: 141516935-141516935 on Build GRCh38
rs771342017 Chr.5: 141516844-141516844 on Build GRCh38
rs755667816 Chr.5: 141516809-141516809 on Build GRCh38
rs901377902 Chr.5: 141516833-141516833 on Build GRCh38
rs375612034 Chr.5: 141516812-141516812 on Build GRCh38
rs1048107293 Chr.5: 141516779-141516779 on Build GRCh38
rs747786198 Chr.5: 141516822-141516827 on Build GRCh38
rs774568872 Chr.5: 141516845-141516845 on Build GRCh38

View Full Product Details