Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75517961-75518234 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1A
Entrez Gene ID
84141
Gene Name
eva-1 homolog A, regulator of programmed cell death
Gene Aliases
FAM176A, TMEM166
UniGene
Hs.302346
Gene Chromosome Location
Chr.2: 75492318-75569722 on Build GRCh38
Transcripts
NM_001135032.1 NM_032181.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5059078 c.83C>T p.S28L Chr.2: 75518058-75518058 on Build GRCh38
1023129 c.23G>A p.S8N Chr.2: 75518118-75518118 on Build GRCh38
5973887 c.66G>A p.A22A Chr.2: 75518075-75518075 on Build GRCh38
3583461 c.75C>T p.S25S Chr.2: 75518066-75518066 on Build GRCh38
2156272 c.67G>T p.A23S Chr.2: 75518074-75518074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754963209 Chr.2: 75518108-75518108 on Build GRCh38
rs890352359 Chr.2: 75518080-75518080 on Build GRCh38
rs775760761 Chr.2: 75518067-75518067 on Build GRCh38
rs766014396 Chr.2: 75518167-75518167 on Build GRCh38
rs762640653 Chr.2: 75518032-75518033 on Build GRCh38
rs369146049 Chr.2: 75518133-75518133 on Build GRCh38
rs532208544 Chr.2: 75518004-75518004 on Build GRCh38
rs769089762 Chr.2: 75518149-75518149 on Build GRCh38
rs777988700 Chr.2: 75518010-75518010 on Build GRCh38
rs780150119 Chr.2: 75518188-75518188 on Build GRCh38
rs369821481 Chr.2: 75518185-75518185 on Build GRCh38
rs1053137113 Chr.2: 75518015-75518015 on Build GRCh38
rs765218766 Chr.2: 75518176-75518176 on Build GRCh38
rs375037470 Chr.2: 75518114-75518114 on Build GRCh38
rs781234792 Chr.2: 75518123-75518123 on Build GRCh38
rs749655008 Chr.2: 75518148-75518148 on Build GRCh38
rs775115947 Chr.2: 75518187-75518187 on Build GRCh38
rs772644493 Chr.2: 75518160-75518160 on Build GRCh38
rs540805520 Chr.2: 75518072-75518072 on Build GRCh38
rs773781691 Chr.2: 75518073-75518073 on Build GRCh38
rs926261338 Chr.2: 75518198-75518200 on Build GRCh38
rs770229887 Chr.2: 75518033-75518033 on Build GRCh38
rs767025615 Chr.2: 75518075-75518075 on Build GRCh38
rs112344664 Chr.2: 75518107-75518107 on Build GRCh38
rs775915837 Chr.2: 75518146-75518146 on Build GRCh38
rs761272832 Chr.2: 75518154-75518154 on Build GRCh38
rs763431830 Chr.2: 75518070-75518070 on Build GRCh38
rs759153258 Chr.2: 75518142-75518142 on Build GRCh38
rs200319399 Chr.2: 75518184-75518184 on Build GRCh38
rs760360094 Chr.2: 75518162-75518162 on Build GRCh38
rs980654901 Chr.2: 75517999-75517999 on Build GRCh38
rs776926428 Chr.2: 75518021-75518021 on Build GRCh38
rs747155416 Chr.2: 75518011-75518011 on Build GRCh38
rs761321988 Chr.2: 75518074-75518074 on Build GRCh38
rs77729933 Chr.2: 75518117-75518117 on Build GRCh38
rs746174348 Chr.2: 75518032-75518032 on Build GRCh38
rs1007492980 Chr.2: 75518082-75518082 on Build GRCh38
rs755761988 Chr.2: 75518103-75518103 on Build GRCh38
rs772689061 Chr.2: 75518028-75518028 on Build GRCh38
rs372051466 Chr.2: 75518111-75518111 on Build GRCh38
rs372483305 Chr.2: 75518172-75518172 on Build GRCh38
rs774857992 Chr.2: 75518151-75518151 on Build GRCh38
rs560233570 Chr.2: 75518076-75518076 on Build GRCh38
rs779001457 Chr.2: 75518110-75518110 on Build GRCh38
rs750530533 Chr.2: 75518186-75518186 on Build GRCh38
rs771211538 Chr.2: 75518016-75518016 on Build GRCh38

View Full Product Details