Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36523148-36523375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3D
Entrez Gene ID
8664
Gene Name
eukaryotic translation initiation factor 3 subunit D
Gene Aliases
EIF3S7, eIF3-p66, eIF3-zeta
UniGene
Hs.55682
Gene Chromosome Location
Chr.22: 36510850-36529431 on Build GRCh38
Transcripts
NM_003753.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1593024 c.492C>A p.A164A Chr.22: 36523182-36523182 on Build GRCh38
6354602 c.422A>G p.Q141R Chr.22: 36523252-36523252 on Build GRCh38
4935593 c.410A>T p.Q137L Chr.22: 36523264-36523264 on Build GRCh38
7104041 c.483G>C p.Q161H Chr.22: 36523191-36523191 on Build GRCh38
6652815 c.506_507delCT p.S169fs*8 Chr.22: 36523167-36523168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751039468 Chr.22: 36523193-36523193 on Build GRCh38
rs62230012 Chr.22: 36523311-36523311 on Build GRCh38
rs140273404 Chr.22: 36523195-36523195 on Build GRCh38
rs8140240 Chr.22: 36523296-36523296 on Build GRCh38
rs767042707 Chr.22: 36523330-36523330 on Build GRCh38
rs1027397689 Chr.22: 36523257-36523257 on Build GRCh38
rs1017781175 Chr.22: 36523307-36523307 on Build GRCh38
rs75384533 Chr.22: 36523313-36523313 on Build GRCh38
rs763602503 Chr.22: 36523183-36523183 on Build GRCh38
rs142365005 Chr.22: 36523240-36523240 on Build GRCh38
rs780955117 Chr.22: 36523329-36523329 on Build GRCh38
rs7290521 Chr.22: 36523347-36523347 on Build GRCh38
rs777903501 Chr.22: 36523276-36523276 on Build GRCh38
rs755293151 Chr.22: 36523236-36523236 on Build GRCh38
rs371251229 Chr.22: 36523291-36523291 on Build GRCh38
rs773901774 Chr.22: 36523317-36523317 on Build GRCh38
rs745832380 Chr.22: 36523299-36523299 on Build GRCh38
rs533562864 Chr.22: 36523260-36523260 on Build GRCh38
rs73169675 Chr.22: 36523328-36523328 on Build GRCh38
rs771590275 Chr.22: 36523298-36523298 on Build GRCh38
rs748339725 Chr.22: 36523263-36523263 on Build GRCh38
rs775163317 Chr.22: 36523310-36523310 on Build GRCh38
rs758550310 Chr.22: 36523266-36523266 on Build GRCh38
rs754182096 Chr.22: 36523232-36523232 on Build GRCh38
rs35723883 Chr.22: 36523346-36523346 on Build GRCh38
rs781192903 Chr.22: 36523301-36523301 on Build GRCh38
rs780599473 Chr.22: 36523212-36523212 on Build GRCh38
rs769664413 Chr.22: 36523306-36523306 on Build GRCh38
rs747116170 Chr.22: 36523282-36523282 on Build GRCh38
rs763724874 Chr.22: 36523314-36523314 on Build GRCh38

View Full Product Details