Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 119690268-119690519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIT
Entrez Gene ID
11113
Gene Name
citron rho-interacting serine/threonine kinase
Gene Aliases
CRIK, STK21
UniGene
Hs.119594
Gene Chromosome Location
Chr.12: 119685790-119877318 on Build GRCh38
Transcripts
NM_001206999.1 NM_007174.2 XM_006719206.2 XM_011537783.1 XM_011537784.1 XM_011537785.1 XM_011537787.1 XM_011537788.1 XM_011537789.1 XM_011537790.2 XM_011537791.2 XM_011537792.2 XM_017018735.1 XM_017018736.1 XM_017018737.1 XM_017018738.1 XM_017018739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476212 c.5903C>T p.P1968L Chr.12: 119690434-119690434 on Build GRCh38
6242517 c.5892C>T p.T1964T Chr.12: 119690319-119690319 on Build GRCh38
2174346 c.5982G>T p.E1994D Chr.12: 119690355-119690355 on Build GRCh38
2174347 c.5856G>T p.E1952D Chr.12: 119690355-119690355 on Build GRCh38
1197044 c.5998C>T p.R2000C Chr.12: 119690339-119690339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751407643 Chr.12: 119690319-119690319 on Build GRCh38
rs201980409 Chr.12: 119690409-119690409 on Build GRCh38
rs139121341 Chr.12: 119690382-119690382 on Build GRCh38
rs758825630 Chr.12: 119690460-119690460 on Build GRCh38
rs768850425 Chr.12: 119690356-119690356 on Build GRCh38
rs772645494 Chr.12: 119690340-119690340 on Build GRCh38
rs368956487 Chr.12: 119690474-119690474 on Build GRCh38
rs373351610 Chr.12: 119690485-119690485 on Build GRCh38
rs751389152 Chr.12: 119690394-119690394 on Build GRCh38
rs766033223 Chr.12: 119690287-119690287 on Build GRCh38
rs754675960 Chr.12: 119690322-119690322 on Build GRCh38
rs866608224 Chr.12: 119690362-119690362 on Build GRCh38
rs1042530265 Chr.12: 119690304-119690304 on Build GRCh38
rs777088999 Chr.12: 119690418-119690418 on Build GRCh38
rs766494281 Chr.12: 119690390-119690390 on Build GRCh38
rs144102132 Chr.12: 119690414-119690414 on Build GRCh38
rs753391537 Chr.12: 119690459-119690459 on Build GRCh38
rs200787337 Chr.12: 119690338-119690338 on Build GRCh38
rs752233553 Chr.12: 119690326-119690326 on Build GRCh38
rs1016583716 Chr.12: 119690291-119690291 on Build GRCh38
rs759454497 Chr.12: 119690402-119690402 on Build GRCh38
rs371376893 Chr.12: 119690359-119690359 on Build GRCh38
rs777373767 Chr.12: 119690331-119690331 on Build GRCh38
rs755620695 Chr.12: 119690328-119690328 on Build GRCh38
rs998721379 Chr.12: 119690343-119690343 on Build GRCh38
rs759661570 Chr.12: 119690488-119690488 on Build GRCh38
rs961048068 Chr.12: 119690333-119690333 on Build GRCh38
rs903012420 Chr.12: 119690316-119690316 on Build GRCh38
rs781051570 Chr.12: 119690324-119690324 on Build GRCh38
rs767249465 Chr.12: 119690403-119690403 on Build GRCh38
rs983828814 Chr.12: 119690453-119690453 on Build GRCh38
rs1027945553 Chr.12: 119690395-119690395 on Build GRCh38
rs560514727 Chr.12: 119690405-119690405 on Build GRCh38
rs368170067 Chr.12: 119690350-119690350 on Build GRCh38
rs769717272 Chr.12: 119690370-119690370 on Build GRCh38
rs755372796 Chr.12: 119690481-119690481 on Build GRCh38
rs761917468 Chr.12: 119690366-119690366 on Build GRCh38
rs780327897 Chr.12: 119690341-119690341 on Build GRCh38
rs928178379 Chr.12: 119690498-119690498 on Build GRCh38
rs762735120 Chr.12: 119690383-119690383 on Build GRCh38
rs780593470 Chr.12: 119690463-119690463 on Build GRCh38
rs766445204 Chr.12: 119690286-119690286 on Build GRCh38

View Full Product Details