Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132547970-132548231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBRSL1
Entrez Gene ID
57666
Gene Name
fibrosin like 1
Gene Aliases
-
UniGene
Hs.411138
Gene Chromosome Location
Chr.12: 132490440-132585188 on Build GRCh38
Transcripts
NM_001142641.1 XM_005266171.3 XM_005266173.3 XM_005266175.3 XM_005266176.3 XM_005266177.3 XM_011534803.2 XM_011534804.2 XM_011534805.2 XM_011534806.2 XM_011534807.2 XM_011534808.2 XM_011534809.2 XM_011534810.2 XM_011534812.2 XM_011534816.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769669 c.644A>G p.K215R Chr.12: 132548031-132548031 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs985903205 Chr.12: 132548195-132548195 on Build GRCh38
rs758472858 Chr.12: 132548023-132548023 on Build GRCh38
rs796389063 Chr.12: 132548166-132548166 on Build GRCh38
rs1031521065 Chr.12: 132548043-132548043 on Build GRCh38
rs184797705 Chr.12: 132548164-132548164 on Build GRCh38
rs1002794537 Chr.12: 132548165-132548165 on Build GRCh38
rs981753353 Chr.12: 132548042-132548042 on Build GRCh38
rs796233396 Chr.12: 132548160-132548160 on Build GRCh38
rs79009737 Chr.12: 132548167-132548167 on Build GRCh38
rs775002573 Chr.12: 132548061-132548061 on Build GRCh38
rs975985761 Chr.12: 132548022-132548022 on Build GRCh38
rs956027994 Chr.12: 132548044-132548044 on Build GRCh38
rs576993256 Chr.12: 132548131-132548131 on Build GRCh38
rs946817998 Chr.12: 132548047-132548047 on Build GRCh38
rs556091453 Chr.12: 132548147-132548147 on Build GRCh38
rs964817015 Chr.12: 132548172-132548172 on Build GRCh38
rs974794149 Chr.12: 132548208-132548208 on Build GRCh38
rs533968864 Chr.12: 132548062-132548062 on Build GRCh38
rs964785904 Chr.12: 132548076-132548076 on Build GRCh38
rs1037828822 Chr.12: 132548148-132548148 on Build GRCh38
rs939618544 Chr.12: 132548090-132548090 on Build GRCh38
rs1014764299 Chr.12: 132547994-132547994 on Build GRCh38
rs932602988 Chr.12: 132548027-132548027 on Build GRCh38
rs540404942 Chr.12: 132548169-132548169 on Build GRCh38
rs914693365 Chr.12: 132548072-132548072 on Build GRCh38
rs762370082 Chr.12: 132548163-132548163 on Build GRCh38
rs149569196 Chr.12: 132548011-132548011 on Build GRCh38
rs539096293 Chr.12: 132548136-132548136 on Build GRCh38
rs1028878897 Chr.12: 132548171-132548171 on Build GRCh38
rs568222638 Chr.12: 132548041-132548041 on Build GRCh38
rs953311269 Chr.12: 132548189-132548189 on Build GRCh38
rs974763103 Chr.12: 132548096-132548096 on Build GRCh38
rs1012603348 Chr.12: 132548156-132548156 on Build GRCh38
rs553958735 Chr.12: 132548102-132548102 on Build GRCh38
rs898290151 Chr.12: 132548152-132548152 on Build GRCh38
rs990240160 Chr.12: 132548053-132548053 on Build GRCh38
rs560247582 Chr.12: 132548210-132548210 on Build GRCh38
rs575871358 Chr.12: 132548153-132548153 on Build GRCh38
rs769083080 Chr.12: 132548026-132548026 on Build GRCh38
rs544844191 Chr.12: 132548162-132548162 on Build GRCh38
rs763212277 Chr.12: 132548019-132548019 on Build GRCh38
rs530592931 Chr.12: 132548050-132548050 on Build GRCh38
rs867539211 Chr.12: 132548012-132548012 on Build GRCh38
rs1011453375 Chr.12: 132548168-132548168 on Build GRCh38
rs1044097371 Chr.12: 132548157-132548157 on Build GRCh38
rs1048424436 Chr.12: 132548132-132548132 on Build GRCh38
rs923249873 Chr.12: 132548103-132548103 on Build GRCh38
rs1042555836 Chr.12: 132548060-132548060 on Build GRCh38

View Full Product Details