Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136833715-136833988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL6
Entrez Gene ID
55684
Gene Name
RAB, member RAS oncogene family-like 6
Gene Aliases
C9orf86, PARF, RBEL1, pp8875
UniGene
Hs.370555
Gene Chromosome Location
Chr.9: 136807922-136841187 on Build GRCh38
Transcripts
NM_001173988.1 NM_001173989.2 NM_024718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551897 c.709G>A p.D237N Chr.9: 136833723-136833723 on Build GRCh38
6013707 c.833G>T p.C278F Chr.9: 136833847-136833847 on Build GRCh38
5799834 c.826C>G p.Q276E Chr.9: 136833840-136833840 on Build GRCh38
6500227 c.877C>T p.P293S Chr.9: 136833891-136833891 on Build GRCh38
6277103 c.721G>T p.A241S Chr.9: 136833735-136833735 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749354825 Chr.9: 136833834-136833834 on Build GRCh38
rs533722396 Chr.9: 136833892-136833892 on Build GRCh38
rs774129497 Chr.9: 136833754-136833754 on Build GRCh38
rs962619648 Chr.9: 136833968-136833968 on Build GRCh38
rs768883190 Chr.9: 136833902-136833902 on Build GRCh38
rs751749870 Chr.9: 136833915-136833915 on Build GRCh38
rs746176397 Chr.9: 136833933-136833933 on Build GRCh38
rs531300465 Chr.9: 136833814-136833814 on Build GRCh38
rs767392980 Chr.9: 136833813-136833813 on Build GRCh38
rs958853678 Chr.9: 136833936-136833936 on Build GRCh38
rs936347810 Chr.9: 136833763-136833763 on Build GRCh38
rs111319027 Chr.9: 136833901-136833901 on Build GRCh38
rs1040888833 Chr.9: 136833827-136833827 on Build GRCh38
rs1014396997 Chr.9: 136833937-136833937 on Build GRCh38
rs774695032 Chr.9: 136833929-136833929 on Build GRCh38
rs775582222 Chr.9: 136833751-136833751 on Build GRCh38
rs1020424389 Chr.9: 136833941-136833942 on Build GRCh38
rs931063313 Chr.9: 136833803-136833803 on Build GRCh38
rs1048217854 Chr.9: 136833805-136833805 on Build GRCh38
rs1014424374 Chr.9: 136833939-136833939 on Build GRCh38
rs760543943 Chr.9: 136833855-136833855 on Build GRCh38
rs918312872 Chr.9: 136833801-136833801 on Build GRCh38
rs1040966214 Chr.9: 136833841-136833841 on Build GRCh38
rs868483714 Chr.9: 136833858-136833858 on Build GRCh38
rs776542500 Chr.9: 136833770-136833772 on Build GRCh38
rs967966996 Chr.9: 136833949-136833949 on Build GRCh38
rs888427455 Chr.9: 136833903-136833903 on Build GRCh38
rs145215909 Chr.9: 136833857-136833857 on Build GRCh38
rs1001970343 Chr.9: 136833907-136833907 on Build GRCh38
rs187468519 Chr.9: 136833750-136833750 on Build GRCh38
rs993034569 Chr.9: 136833741-136833741 on Build GRCh38
rs137939933 Chr.9: 136833958-136833958 on Build GRCh38
rs757522707 Chr.9: 136833919-136833919 on Build GRCh38
rs761628258 Chr.9: 136833759-136833759 on Build GRCh38
rs781414397 Chr.9: 136833922-136833922 on Build GRCh38
rs972826736 Chr.9: 136833785-136833785 on Build GRCh38
rs758351837 Chr.9: 136833879-136833879 on Build GRCh38
rs901104991 Chr.9: 136833848-136833848 on Build GRCh38
rs970398840 Chr.9: 136833758-136833758 on Build GRCh38
rs904569706 Chr.9: 136833874-136833874 on Build GRCh38
rs376102803 Chr.9: 136833856-136833856 on Build GRCh38

View Full Product Details