Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130762093-130762366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMER3
Entrez Gene ID
205147
Gene Name
APC membrane recruitment protein 3
Gene Aliases
FAM123C
UniGene
Hs.369289
Gene Chromosome Location
Chr.2: 130754475-130768134 on Build GRCh38
Transcripts
NM_001105193.1 NM_001105194.1 NM_001105195.1 NM_152698.2 XM_011510793.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4965174 c.186C>T p.Y62Y Chr.2: 130762258-130762258 on Build GRCh38
715805 c.105C>A p.G35G Chr.2: 130762177-130762177 on Build GRCh38
3836830 c.181G>T p.E61* Chr.2: 130762253-130762253 on Build GRCh38
4532794 c.191G>A p.R64K Chr.2: 130762263-130762263 on Build GRCh38
6087607 c.82G>T p.A28S Chr.2: 130762154-130762154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764257062 Chr.2: 130762190-130762190 on Build GRCh38
rs772482449 Chr.2: 130762141-130762141 on Build GRCh38
rs996368182 Chr.2: 130762179-130762179 on Build GRCh38
rs780560291 Chr.2: 130762224-130762224 on Build GRCh38
rs371220044 Chr.2: 130762259-130762259 on Build GRCh38
rs551831783 Chr.2: 130762306-130762306 on Build GRCh38
rs191484242 Chr.2: 130762176-130762176 on Build GRCh38
rs755353161 Chr.2: 130762312-130762312 on Build GRCh38
rs762376021 Chr.2: 130762152-130762152 on Build GRCh38
rs996077726 Chr.2: 130762133-130762133 on Build GRCh38
rs780428758 Chr.2: 130762307-130762307 on Build GRCh38
rs370830797 Chr.2: 130762313-130762313 on Build GRCh38
rs770690056 Chr.2: 130762326-130762326 on Build GRCh38
rs372997529 Chr.2: 130762203-130762203 on Build GRCh38
rs769354675 Chr.2: 130762150-130762150 on Build GRCh38
rs768996309 Chr.2: 130762237-130762237 on Build GRCh38
rs1018048222 Chr.2: 130762163-130762163 on Build GRCh38
rs374790815 Chr.2: 130762303-130762303 on Build GRCh38
rs765350144 Chr.2: 130762301-130762301 on Build GRCh38
rs749980804 Chr.2: 130762334-130762334 on Build GRCh38
rs746316318 Chr.2: 130762134-130762134 on Build GRCh38
rs374506642 Chr.2: 130762151-130762151 on Build GRCh38
rs750523251 Chr.2: 130762302-130762302 on Build GRCh38
rs919762235 Chr.2: 130762273-130762273 on Build GRCh38
rs749347187 Chr.2: 130762126-130762126 on Build GRCh38
rs754066461 Chr.2: 130762293-130762293 on Build GRCh38
rs929759050 Chr.2: 130762295-130762295 on Build GRCh38
rs151222824 Chr.2: 130762123-130762123 on Build GRCh38
rs374950545 Chr.2: 130762241-130762241 on Build GRCh38
rs771546193 Chr.2: 130762336-130762336 on Build GRCh38
rs770510481 Chr.2: 130762153-130762153 on Build GRCh38
rs763893750 Chr.2: 130762287-130762287 on Build GRCh38
rs753033858 Chr.2: 130762114-130762114 on Build GRCh38
rs140913895 Chr.2: 130762158-130762158 on Build GRCh38
rs917548134 Chr.2: 130762317-130762317 on Build GRCh38
rs775800844 Chr.2: 130762143-130762143 on Build GRCh38
rs973917344 Chr.2: 130762265-130762265 on Build GRCh38
rs773964038 Chr.2: 130762280-130762280 on Build GRCh38
rs762018995 Chr.2: 130762298-130762298 on Build GRCh38
rs138837389 Chr.2: 130762169-130762169 on Build GRCh38
rs775305041 Chr.2: 130762341-130762341 on Build GRCh38
rs777954595 Chr.2: 130762124-130762124 on Build GRCh38
rs747363525 Chr.2: 130762227-130762227 on Build GRCh38
rs757285578 Chr.2: 130762127-130762127 on Build GRCh38
rs752044425 Chr.2: 130762311-130762311 on Build GRCh38
rs779395303 Chr.2: 130762132-130762132 on Build GRCh38
rs1019605491 Chr.2: 130762167-130762167 on Build GRCh38
rs549136165 Chr.2: 130762199-130762199 on Build GRCh38
rs767464409 Chr.2: 130762173-130762173 on Build GRCh38
rs778664237 Chr.2: 130762335-130762335 on Build GRCh38
rs979659922 Chr.2: 130762233-130762233 on Build GRCh38
rs1007873530 Chr.2: 130762154-130762154 on Build GRCh38
rs746954182 Chr.2: 130762345-130762345 on Build GRCh38
rs754027088 Chr.2: 130762193-130762193 on Build GRCh38
rs369336031 Chr.2: 130762258-130762258 on Build GRCh38
rs750996134 Chr.2: 130762201-130762201 on Build GRCh38
rs747885162 Chr.2: 130762145-130762145 on Build GRCh38
rs376219909 Chr.2: 130762282-130762282 on Build GRCh38
rs200636300 Chr.2: 130762281-130762281 on Build GRCh38
rs781612485 Chr.2: 130762318-130762318 on Build GRCh38
rs897382592 Chr.2: 130762337-130762337 on Build GRCh38
rs573327367 Chr.2: 130762325-130762325 on Build GRCh38
rs764765629 Chr.2: 130762174-130762175 on Build GRCh38

View Full Product Details