Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 19192252-19192517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP3
Entrez Gene ID
8048
Gene Name
cysteine and glycine rich protein 3
Gene Aliases
CLP, CMD1M, CMH12, CRP3, LMO4, MLP
UniGene
Hs.83577
Gene Chromosome Location
Chr.11: 19182030-19210571 on Build GRCh38
Transcripts
NM_003476.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1353126 c.64G>T p.E22* Chr.11: 19192385-19192385 on Build GRCh38
4559557 c.80G>A p.G27E Chr.11: 19192369-19192369 on Build GRCh38
3782604 c.99G>A p.T33T Chr.11: 19192350-19192350 on Build GRCh38
6483748 c.48C>A p.T16T Chr.11: 19192401-19192401 on Build GRCh38
466656 c.36C>T p.A12A Chr.11: 19192413-19192413 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766395852 Chr.11: 19192377-19192377 on Build GRCh38
rs397516857 Chr.11: 19192402-19192402 on Build GRCh38
rs147549410 Chr.11: 19192350-19192350 on Build GRCh38
rs762730416 Chr.11: 19192444-19192444 on Build GRCh38
rs933469401 Chr.11: 19192484-19192484 on Build GRCh38
rs759305806 Chr.11: 19192397-19192397 on Build GRCh38
rs776468900 Chr.11: 19192340-19192340 on Build GRCh38
rs769677095 Chr.11: 19192436-19192436 on Build GRCh38
rs935934212 Chr.11: 19192440-19192440 on Build GRCh38
rs185421701 Chr.11: 19192298-19192298 on Build GRCh38
rs151177504 Chr.11: 19192428-19192428 on Build GRCh38
rs774006811 Chr.11: 19192459-19192459 on Build GRCh38
rs775270112 Chr.11: 19192331-19192331 on Build GRCh38
rs753120437 Chr.11: 19192401-19192401 on Build GRCh38
rs758947977 Chr.11: 19192351-19192351 on Build GRCh38
rs774314682 Chr.11: 19192312-19192312 on Build GRCh38
rs776444956 Chr.11: 19192394-19192400 on Build GRCh38
rs547580470 Chr.11: 19192291-19192291 on Build GRCh38
rs372446591 Chr.11: 19192480-19192480 on Build GRCh38
rs863225265 Chr.11: 19192356-19192356 on Build GRCh38
rs768173909 Chr.11: 19192469-19192469 on Build GRCh38
rs768436996 Chr.11: 19192337-19192337 on Build GRCh38
rs768595166 Chr.11: 19192429-19192429 on Build GRCh38
rs766815585 Chr.11: 19192300-19192300 on Build GRCh38
rs372717179 Chr.11: 19192363-19192363 on Build GRCh38
rs182726860 Chr.11: 19192411-19192411 on Build GRCh38
rs758897446 Chr.11: 19192299-19192299 on Build GRCh38
rs777847267 Chr.11: 19192321-19192321 on Build GRCh38
rs45550635 Chr.11: 19192439-19192439 on Build GRCh38
rs112513407 Chr.11: 19192479-19192479 on Build GRCh38
rs185980145 Chr.11: 19192433-19192433 on Build GRCh38
rs760384315 Chr.11: 19192451-19192451 on Build GRCh38
rs375525973 Chr.11: 19192394-19192394 on Build GRCh38
rs763193066 Chr.11: 19192341-19192341 on Build GRCh38
rs746990436 Chr.11: 19192336-19192336 on Build GRCh38
rs45531937 Chr.11: 19192427-19192427 on Build GRCh38
rs777883490 Chr.11: 19192409-19192409 on Build GRCh38
rs869025378 Chr.11: 19192400-19192400 on Build GRCh38
rs369736950 Chr.11: 19192313-19192313 on Build GRCh38
rs756414072 Chr.11: 19192320-19192320 on Build GRCh38
rs767932680 Chr.11: 19192400-19192400 on Build GRCh38
rs781731204 Chr.11: 19192318-19192318 on Build GRCh38
rs889450443 Chr.11: 19192278-19192278 on Build GRCh38
rs748363797 Chr.11: 19192319-19192319 on Build GRCh38
rs773319934 Chr.11: 19192318-19192318 on Build GRCh38
rs752154978 Chr.11: 19192308-19192308 on Build GRCh38
rs1053281170 Chr.11: 19192485-19192485 on Build GRCh38
rs190182843 Chr.11: 19192375-19192375 on Build GRCh38
rs909246685 Chr.11: 19192371-19192371 on Build GRCh38
rs760600548 Chr.11: 19192339-19192339 on Build GRCh38
rs765951170 Chr.11: 19192364-19192364 on Build GRCh38
rs992234249 Chr.11: 19192406-19192406 on Build GRCh38
rs747959696 Chr.11: 19192434-19192434 on Build GRCh38
rs772727223 Chr.11: 19192359-19192359 on Build GRCh38
rs760049800 Chr.11: 19192463-19192463 on Build GRCh38
rs756368977 Chr.11: 19192407-19192407 on Build GRCh38
rs749361872 Chr.11: 19192326-19192326 on Build GRCh38
rs141980088 Chr.11: 19192420-19192420 on Build GRCh38
rs772000025 Chr.11: 19192327-19192327 on Build GRCh38

View Full Product Details