Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45871897-45872158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA3
Entrez Gene ID
3171
Gene Name
forkhead box A3
Gene Aliases
FKHH3, HNF3G, TCF3G
UniGene
Hs.36137
Gene Chromosome Location
Chr.19: 45864260-45873797 on Build GRCh38
Transcripts
NM_004497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672951 c.109C>A p.L37I Chr.19: 45872114-45872114 on Build GRCh38
5390117 c.133C>T p.P45S Chr.19: 45872138-45872138 on Build GRCh38
1726131 c.131A>G p.N44S Chr.19: 45872136-45872136 on Build GRCh38
6323706 c.87C>T p.T29T Chr.19: 45872092-45872092 on Build GRCh38
4127723 c.152C>A p.P51H Chr.19: 45872157-45872157 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759247371 Chr.19: 45872048-45872048 on Build GRCh38
rs764471778 Chr.19: 45872074-45872074 on Build GRCh38
rs891423347 Chr.19: 45871943-45871943 on Build GRCh38
rs780122523 Chr.19: 45872085-45872085 on Build GRCh38
rs567465651 Chr.19: 45871952-45871952 on Build GRCh38
rs367990040 Chr.19: 45872007-45872007 on Build GRCh38
rs770728557 Chr.19: 45872043-45872043 on Build GRCh38
rs754343671 Chr.19: 45872078-45872078 on Build GRCh38
rs376849431 Chr.19: 45872095-45872095 on Build GRCh38
rs201518711 Chr.19: 45872058-45872058 on Build GRCh38
rs143578664 Chr.19: 45872086-45872086 on Build GRCh38
rs773081512 Chr.19: 45872027-45872027 on Build GRCh38
rs752304015 Chr.19: 45872083-45872083 on Build GRCh38
rs759303786 Chr.19: 45871969-45871969 on Build GRCh38
rs762614607 Chr.19: 45872041-45872041 on Build GRCh38
rs566369055 Chr.19: 45872106-45872106 on Build GRCh38
rs1052790786 Chr.19: 45871921-45871921 on Build GRCh38
rs757615875 Chr.19: 45872079-45872079 on Build GRCh38
rs975000288 Chr.19: 45871999-45871999 on Build GRCh38
rs1011361734 Chr.19: 45871956-45871960 on Build GRCh38
rs199885627 Chr.19: 45872069-45872069 on Build GRCh38
rs779323164 Chr.19: 45872082-45872082 on Build GRCh38
rs550382199 Chr.19: 45872044-45872044 on Build GRCh38
rs757817912 Chr.19: 45872131-45872131 on Build GRCh38
rs749227057 Chr.19: 45872090-45872090 on Build GRCh38
rs979170792 Chr.19: 45871941-45871941 on Build GRCh38
rs570167287 Chr.19: 45872054-45872054 on Build GRCh38
rs984261233 Chr.19: 45872104-45872104 on Build GRCh38
rs771789911 Chr.19: 45872128-45872128 on Build GRCh38
rs758551287 Chr.19: 45872084-45872084 on Build GRCh38
rs764786295 Chr.19: 45871995-45871995 on Build GRCh38
rs966892712 Chr.19: 45871927-45871927 on Build GRCh38
rs759264087 Chr.19: 45872116-45872116 on Build GRCh38
rs201137445 Chr.19: 45872087-45872087 on Build GRCh38

View Full Product Details