Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 141151582-141151853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND3
Entrez Gene ID
22898
Gene Name
DENN domain containing 3
Gene Aliases
-
UniGene
Hs.18166
Gene Chromosome Location
Chr.8: 141128618-141195804 on Build GRCh38
Transcripts
NM_014957.2 XM_005250838.4 XM_005250839.2 XM_005250840.4 XM_005250841.3 XM_005250842.4 XM_011516933.1 XM_011516934.2 XM_011516937.1 XM_011516941.2 XM_017013241.1 XM_017013242.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1132828 c.662C>A p.S221* Chr.8: 141151665-141151665 on Build GRCh38
454103 c.778G>A p.A260T Chr.8: 141151781-141151781 on Build GRCh38
3924732 c.657C>T p.F219F Chr.8: 141151660-141151660 on Build GRCh38
3645778 c.738C>T p.F246F Chr.8: 141151741-141151741 on Build GRCh38
6266574 c.644G>T p.R215L Chr.8: 141151647-141151647 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767488832 Chr.8: 141151755-141151755 on Build GRCh38
rs1037631038 Chr.8: 141151745-141151745 on Build GRCh38
rs778157810 Chr.8: 141151822-141151822 on Build GRCh38
rs778661680 Chr.8: 141151697-141151697 on Build GRCh38
rs375291304 Chr.8: 141151603-141151603 on Build GRCh38
rs747569316 Chr.8: 141151733-141151733 on Build GRCh38
rs775793114 Chr.8: 141151751-141151751 on Build GRCh38
rs772531286 Chr.8: 141151643-141151643 on Build GRCh38
rs377138571 Chr.8: 141151684-141151684 on Build GRCh38
rs368117361 Chr.8: 141151638-141151638 on Build GRCh38
rs770972671 Chr.8: 141151738-141151738 on Build GRCh38
rs77142252 Chr.8: 141151720-141151720 on Build GRCh38
rs758744672 Chr.8: 141151814-141151814 on Build GRCh38
rs138968166 Chr.8: 141151687-141151687 on Build GRCh38
rs1033499486 Chr.8: 141151615-141151615 on Build GRCh38
rs769858175 Chr.8: 141151748-141151748 on Build GRCh38
rs752026665 Chr.8: 141151823-141151823 on Build GRCh38
rs542351463 Chr.8: 141151628-141151628 on Build GRCh38
rs542335448 Chr.8: 141151665-141151665 on Build GRCh38
rs748961763 Chr.8: 141151616-141151616 on Build GRCh38
rs2304283 Chr.8: 141151789-141151789 on Build GRCh38
rs755358164 Chr.8: 141151631-141151631 on Build GRCh38
rs200254442 Chr.8: 141151611-141151611 on Build GRCh38
rs755700994 Chr.8: 141151607-141151607 on Build GRCh38
rs757181115 Chr.8: 141151604-141151604 on Build GRCh38
rs2304284 Chr.8: 141151774-141151774 on Build GRCh38
rs531198735 Chr.8: 141151741-141151741 on Build GRCh38
rs781307331 Chr.8: 141151605-141151605 on Build GRCh38
rs761636095 Chr.8: 141151753-141151753 on Build GRCh38
rs773643758 Chr.8: 141151669-141151670 on Build GRCh38
rs773734634 Chr.8: 141151647-141151647 on Build GRCh38
rs201883569 Chr.8: 141151756-141151756 on Build GRCh38
rs776816199 Chr.8: 141151655-141151655 on Build GRCh38
rs753510043 Chr.8: 141151691-141151691 on Build GRCh38
rs368235736 Chr.8: 141151766-141151766 on Build GRCh38
rs373705581 Chr.8: 141151651-141151651 on Build GRCh38
rs773977058 Chr.8: 141151666-141151666 on Build GRCh38
rs758763055 Chr.8: 141151719-141151719 on Build GRCh38
rs767503171 Chr.8: 141151683-141151683 on Build GRCh38
rs150268914 Chr.8: 141151676-141151676 on Build GRCh38
rs765959967 Chr.8: 141151775-141151775 on Build GRCh38
rs571831372 Chr.8: 141151652-141151652 on Build GRCh38
rs755680824 Chr.8: 141151685-141151685 on Build GRCh38
rs142459324 Chr.8: 141151813-141151813 on Build GRCh38
rs754630590 Chr.8: 141151692-141151692 on Build GRCh38
rs376487961 Chr.8: 141151654-141151654 on Build GRCh38
rs745990732 Chr.8: 141151742-141151742 on Build GRCh38
rs928858016 Chr.8: 141151793-141151793 on Build GRCh38
rs371991598 Chr.8: 141151639-141151639 on Build GRCh38
rs753251817 Chr.8: 141151780-141151780 on Build GRCh38
rs977526362 Chr.8: 141151660-141151660 on Build GRCh38
rs757719239 Chr.8: 141151829-141151829 on Build GRCh38
rs748416043 Chr.8: 141151714-141151714 on Build GRCh38
rs772415482 Chr.8: 141151653-141151655 on Build GRCh38
rs754541615 Chr.8: 141151788-141151788 on Build GRCh38
rs910402732 Chr.8: 141151707-141151707 on Build GRCh38
rs759722612 Chr.8: 141151662-141151662 on Build GRCh38

View Full Product Details