Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44710738-44711011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPDA2
Entrez Gene ID
132789
Gene Name
glucosamine-6-phosphate deaminase 2
Gene Aliases
GNP2, SB52
UniGene
Hs.21398
Gene Chromosome Location
Chr.4: 44701795-44726634 on Build GRCh38
Transcripts
NM_001270880.1 NM_001270881.1 NM_138335.2 NR_073094.1 NR_073095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1055634 c.554C>A p.A185D Chr.4: 44710993-44710993 on Build GRCh38
4124863 c.562G>A p.V188I Chr.4: 44710985-44710985 on Build GRCh38
1618860 c.549T>G p.T183T Chr.4: 44710998-44710998 on Build GRCh38
6857485 c.564T>G p.V188V Chr.4: 44710983-44710983 on Build GRCh38
734383 c.536C>G p.S179* Chr.4: 44711011-44711011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775500121 Chr.4: 44710932-44710932 on Build GRCh38
rs190590795 Chr.4: 44710903-44710903 on Build GRCh38
rs566447075 Chr.4: 44710877-44710877 on Build GRCh38
rs773133423 Chr.4: 44710951-44710951 on Build GRCh38
rs569133200 Chr.4: 44710810-44710810 on Build GRCh38
rs368488482 Chr.4: 44710927-44710927 on Build GRCh38
rs989382842 Chr.4: 44710816-44710816 on Build GRCh38
rs576567597 Chr.4: 44710926-44710926 on Build GRCh38
rs772654925 Chr.4: 44710914-44710914 on Build GRCh38
rs373590544 Chr.4: 44710870-44710870 on Build GRCh38
rs770088618 Chr.4: 44710930-44710930 on Build GRCh38
rs781733548 Chr.4: 44710916-44710916 on Build GRCh38
rs762998394 Chr.4: 44710936-44710936 on Build GRCh38
rs556338548 Chr.4: 44710981-44710981 on Build GRCh38
rs746012393 Chr.4: 44710929-44710929 on Build GRCh38
rs748494433 Chr.4: 44710926-44710929 on Build GRCh38
rs770473660 Chr.4: 44710929-44710932 on Build GRCh38
rs778400882 Chr.4: 44710917-44710917 on Build GRCh38
rs913847247 Chr.4: 44710836-44710836 on Build GRCh38
rs749391536 Chr.4: 44710982-44710984 on Build GRCh38
rs115053108 Chr.4: 44710973-44710973 on Build GRCh38
rs936351017 Chr.4: 44710802-44710802 on Build GRCh38
rs975479302 Chr.4: 44710764-44710764 on Build GRCh38
rs947453118 Chr.4: 44710852-44710852 on Build GRCh38
rs989326312 Chr.4: 44710796-44710796 on Build GRCh38
rs3749582 Chr.4: 44710891-44710891 on Build GRCh38
rs977415963 Chr.4: 44710859-44710859 on Build GRCh38
rs143143289 Chr.4: 44710953-44710953 on Build GRCh38
rs536553686 Chr.4: 44710828-44710828 on Build GRCh38
rs370380122 Chr.4: 44710950-44710950 on Build GRCh38
rs928696945 Chr.4: 44710895-44710895 on Build GRCh38
rs778093288 Chr.4: 44710939-44710942 on Build GRCh38
rs945172245 Chr.4: 44710871-44710871 on Build GRCh38
rs372340439 Chr.4: 44710939-44710939 on Build GRCh38
rs550835688 Chr.4: 44710779-44710779 on Build GRCh38
rs750120090 Chr.4: 44710925-44710925 on Build GRCh38

View Full Product Details