Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108899298-108899571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAO
Entrez Gene ID
1610
Gene Name
D-amino acid oxidase
Gene Aliases
DAAO, DAMOX, OXDA
UniGene
Hs.113227
Gene Chromosome Location
Chr.12: 108880073-108901043 on Build GRCh38
Transcripts
NM_001917.4 XM_005268692.3 XM_011538004.2 XM_011538005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160327 c.814-1G>C p.? Chr.12: 108899376-108899376 on Build GRCh38
280290 c.868C>T p.R290W Chr.12: 108899431-108899431 on Build GRCh38
934658 c.904A>C p.N302H Chr.12: 108899467-108899467 on Build GRCh38
310471 c.870G>T p.R290R Chr.12: 108899433-108899433 on Build GRCh38
2154560 c.847C>T p.R283W Chr.12: 108899410-108899410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764531224 Chr.12: 108899383-108899383 on Build GRCh38
rs143550642 Chr.12: 108899411-108899411 on Build GRCh38
rs3918347 Chr.12: 108899544-108899544 on Build GRCh38
rs772094370 Chr.12: 108899403-108899403 on Build GRCh38
rs756775362 Chr.12: 108899381-108899381 on Build GRCh38
rs888461072 Chr.12: 108899399-108899399 on Build GRCh38
rs771014745 Chr.12: 108899428-108899428 on Build GRCh38
rs775976658 Chr.12: 108899440-108899440 on Build GRCh38
rs371256077 Chr.12: 108899471-108899471 on Build GRCh38
rs140015394 Chr.12: 108899410-108899410 on Build GRCh38
rs756480670 Chr.12: 108899348-108899348 on Build GRCh38
rs761019624 Chr.12: 108899445-108899445 on Build GRCh38
rs146524799 Chr.12: 108899400-108899400 on Build GRCh38
rs746734558 Chr.12: 108899414-108899414 on Build GRCh38
rs1009795554 Chr.12: 108899506-108899506 on Build GRCh38
rs930865044 Chr.12: 108899369-108899369 on Build GRCh38
rs753015643 Chr.12: 108899346-108899346 on Build GRCh38
rs11420034 Chr.12: 108899333-108899334 on Build GRCh38
rs570072379 Chr.12: 108899343-108899343 on Build GRCh38
rs201364697 Chr.12: 108899431-108899431 on Build GRCh38
rs754602852 Chr.12: 108899334-108899334 on Build GRCh38
rs372560295 Chr.12: 108899432-108899432 on Build GRCh38
rs113854555 Chr.12: 108899407-108899407 on Build GRCh38
rs752161005 Chr.12: 108899491-108899491 on Build GRCh38
rs375177190 Chr.12: 108899352-108899352 on Build GRCh38
rs398116858 Chr.12: 108899342-108899343 on Build GRCh38
rs763136408 Chr.12: 108899424-108899424 on Build GRCh38
rs565608617 Chr.12: 108899337-108899337 on Build GRCh38
rs766429341 Chr.12: 108899326-108899326 on Build GRCh38
rs566264618 Chr.12: 108899420-108899420 on Build GRCh38
rs774565696 Chr.12: 108899525-108899525 on Build GRCh38
rs777413514 Chr.12: 108899355-108899355 on Build GRCh38
rs73410968 Chr.12: 108899360-108899360 on Build GRCh38
rs532675157 Chr.12: 108899345-108899345 on Build GRCh38
rs537805560 Chr.12: 108899510-108899510 on Build GRCh38
rs747000236 Chr.12: 108899490-108899490 on Build GRCh38
rs756901160 Chr.12: 108899449-108899449 on Build GRCh38
rs748004248 Chr.12: 108899511-108899511 on Build GRCh38
rs760079225 Chr.12: 108899338-108899338 on Build GRCh38
rs778334098 Chr.12: 108899390-108899390 on Build GRCh38
rs771006214 Chr.12: 108899513-108899513 on Build GRCh38
rs745766982 Chr.12: 108899398-108899398 on Build GRCh38
rs764973777 Chr.12: 108899452-108899452 on Build GRCh38
rs760432003 Chr.12: 108899453-108899453 on Build GRCh38
rs75885639 Chr.12: 108899507-108899507 on Build GRCh38
rs146793573 Chr.12: 108899430-108899430 on Build GRCh38
rs764420954 Chr.12: 108899446-108899446 on Build GRCh38
rs768403852 Chr.12: 108899419-108899419 on Build GRCh38
rs376433124 Chr.12: 108899456-108899456 on Build GRCh38
rs929207744 Chr.12: 108899372-108899372 on Build GRCh38

View Full Product Details