Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10901401-10901658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIITA
Entrez Gene ID
4261
Gene Name
class II, major histocompatibility complex, transactivator
Gene Aliases
C2TA, CIITAIV, MHC2TA, NLRA
UniGene
Hs.592051
Gene Chromosome Location
Chr.16: 10866208-10941562 on Build GRCh38
Transcripts
NM_000246.3 NM_001286402.1 NM_001286403.1 NR_104444.1 XM_006720880.3 XM_011522484.2 XM_011522485.2 XM_011522486.2 XM_011522487.2 XM_011522488.2 XM_011522489.2 XM_011522490.2 XM_011522491.2 XM_011522492.2 XM_011522493.2 XM_011522494.2 XM_011522495.2 XM_011522496.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3505730 c.447G>A p.E149E Chr.16: 10901524-10901524 on Build GRCh38
7347648 c.437-6C>A p.? Chr.16: 10901508-10901508 on Build GRCh38
1741257 c.455C>T p.P152L Chr.16: 10901532-10901532 on Build GRCh38
434599 c.460G>C p.D154H Chr.16: 10901537-10901537 on Build GRCh38
6375907 c.473G>T p.W158L Chr.16: 10901550-10901550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200780683 Chr.16: 10901510-10901510 on Build GRCh38
rs910534495 Chr.16: 10901524-10901524 on Build GRCh38
rs764794681 Chr.16: 10901585-10901589 on Build GRCh38
rs770295658 Chr.16: 10901514-10901514 on Build GRCh38
rs758108252 Chr.16: 10901560-10901560 on Build GRCh38
rs778625872 Chr.16: 10901470-10901470 on Build GRCh38
rs767774238 Chr.16: 10901464-10901464 on Build GRCh38
rs908964514 Chr.16: 10901636-10901636 on Build GRCh38
rs774695473 Chr.16: 10901532-10901532 on Build GRCh38
rs534049594 Chr.16: 10901449-10901449 on Build GRCh38
rs754557219 Chr.16: 10901595-10901595 on Build GRCh38
rs147666647 Chr.16: 10901553-10901553 on Build GRCh38
rs779268248 Chr.16: 10901450-10901450 on Build GRCh38
rs1021661953 Chr.16: 10901458-10901458 on Build GRCh38
rs776453613 Chr.16: 10901507-10901507 on Build GRCh38
rs758428305 Chr.16: 10901487-10901487 on Build GRCh38
rs998822239 Chr.16: 10901478-10901478 on Build GRCh38
rs763218055 Chr.16: 10901527-10901527 on Build GRCh38
rs149990869 Chr.16: 10901548-10901548 on Build GRCh38
rs373022799 Chr.16: 10901529-10901529 on Build GRCh38
rs751411095 Chr.16: 10901583-10901583 on Build GRCh38
rs904266388 Chr.16: 10901572-10901572 on Build GRCh38
rs944728823 Chr.16: 10901525-10901525 on Build GRCh38
rs116778555 Chr.16: 10901609-10901609 on Build GRCh38
rs986229740 Chr.16: 10901499-10901499 on Build GRCh38
rs779844104 Chr.16: 10901488-10901488 on Build GRCh38
rs765700885 Chr.16: 10901474-10901474 on Build GRCh38
rs534970350 Chr.16: 10901556-10901556 on Build GRCh38
rs760823592 Chr.16: 10901539-10901539 on Build GRCh38
rs962178962 Chr.16: 10901446-10901446 on Build GRCh38
rs151317882 Chr.16: 10901533-10901533 on Build GRCh38
rs565948365 Chr.16: 10901515-10901515 on Build GRCh38
rs780081931 Chr.16: 10901565-10901565 on Build GRCh38
rs76923280 Chr.16: 10901537-10901537 on Build GRCh38
rs113282345 Chr.16: 10901455-10901455 on Build GRCh38
rs767539454 Chr.16: 10901534-10901534 on Build GRCh38
rs757069078 Chr.16: 10901466-10901466 on Build GRCh38
rs45543731 Chr.16: 10901623-10901623 on Build GRCh38
rs930744370 Chr.16: 10901497-10901497 on Build GRCh38
rs546862746 Chr.16: 10901432-10901432 on Build GRCh38
rs773558566 Chr.16: 10901523-10901523 on Build GRCh38
rs745802386 Chr.16: 10901481-10901481 on Build GRCh38
rs768251799 Chr.16: 10901503-10901503 on Build GRCh38
rs746579744 Chr.16: 10901490-10901490 on Build GRCh38
rs999872773 Chr.16: 10901579-10901580 on Build GRCh38

View Full Product Details