Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15010916-15011173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC105
Entrez Gene ID
126402
Gene Name
coiled-coil domain containing 105
Gene Aliases
-
UniGene
Hs.375985
Gene Chromosome Location
Chr.19: 15010727-15023271 on Build GRCh38
Transcripts
NM_173482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1564650 c.239G>T p.G80V Chr.19: 15011064-15011064 on Build GRCh38
6423558 c.254A>G p.E85G Chr.19: 15011079-15011079 on Build GRCh38
5639217 c.214C>A p.R72S Chr.19: 15011039-15011039 on Build GRCh38
6408680 c.313T>C p.W105R Chr.19: 15011138-15011138 on Build GRCh38
1391011 c.105G>A p.L35L Chr.19: 15010930-15010930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77764676 Chr.19: 15010997-15010997 on Build GRCh38
rs773331192 Chr.19: 15011136-15011136 on Build GRCh38
rs756715181 Chr.19: 15010973-15010973 on Build GRCh38
rs545556924 Chr.19: 15011154-15011154 on Build GRCh38
rs780712831 Chr.19: 15011065-15011065 on Build GRCh38
rs774261249 Chr.19: 15011009-15011009 on Build GRCh38
rs1055960767 Chr.19: 15011006-15011006 on Build GRCh38
rs578094489 Chr.19: 15011099-15011099 on Build GRCh38
rs754428766 Chr.19: 15011048-15011048 on Build GRCh38
rs772136963 Chr.19: 15011119-15011119 on Build GRCh38
rs150456275 Chr.19: 15010933-15010933 on Build GRCh38
rs199808866 Chr.19: 15011148-15011148 on Build GRCh38
rs200957520 Chr.19: 15011095-15011095 on Build GRCh38
rs767960145 Chr.19: 15010941-15010941 on Build GRCh38
rs980046072 Chr.19: 15010995-15010995 on Build GRCh38
rs866949633 Chr.19: 15011106-15011106 on Build GRCh38
rs1045025463 Chr.19: 15011054-15011054 on Build GRCh38
rs772720021 Chr.19: 15010943-15010943 on Build GRCh38
rs568806898 Chr.19: 15011084-15011084 on Build GRCh38
rs781099838 Chr.19: 15010994-15010994 on Build GRCh38
rs751267551 Chr.19: 15010988-15010988 on Build GRCh38
rs903431734 Chr.19: 15011074-15011074 on Build GRCh38
rs999075824 Chr.19: 15011077-15011077 on Build GRCh38
rs372938359 Chr.19: 15010978-15010978 on Build GRCh38
rs777380028 Chr.19: 15010982-15010982 on Build GRCh38
rs760962966 Chr.19: 15011040-15011040 on Build GRCh38
rs760942873 Chr.19: 15011144-15011144 on Build GRCh38
rs779103514 Chr.19: 15011092-15011092 on Build GRCh38
rs779977866 Chr.19: 15011005-15011005 on Build GRCh38
rs116070180 Chr.19: 15011046-15011046 on Build GRCh38
rs768752309 Chr.19: 15011008-15011008 on Build GRCh38
rs927242256 Chr.19: 15011061-15011061 on Build GRCh38
rs750032164 Chr.19: 15011072-15011072 on Build GRCh38
rs773421461 Chr.19: 15011025-15011025 on Build GRCh38
rs975723757 Chr.19: 15010951-15010951 on Build GRCh38
rs761899374 Chr.19: 15010934-15010934 on Build GRCh38
rs909726800 Chr.19: 15010996-15010996 on Build GRCh38
rs772183332 Chr.19: 15011020-15011020 on Build GRCh38
rs750727974 Chr.19: 15010942-15010942 on Build GRCh38
rs757000109 Chr.19: 15011064-15011064 on Build GRCh38
rs142952980 Chr.19: 15011012-15011012 on Build GRCh38
rs201312674 Chr.19: 15011088-15011088 on Build GRCh38
rs756837621 Chr.19: 15010991-15010991 on Build GRCh38
rs751020384 Chr.19: 15011053-15011053 on Build GRCh38
rs568010037 Chr.19: 15011073-15011073 on Build GRCh38
rs555673221 Chr.19: 15011050-15011050 on Build GRCh38
rs767002210 Chr.19: 15010974-15010974 on Build GRCh38
rs749173220 Chr.19: 15011007-15011007 on Build GRCh38
rs138297206 Chr.19: 15010981-15010981 on Build GRCh38

View Full Product Details