Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71617071-71617296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR3
Entrez Gene ID
2833
Gene Name
C-X-C motif chemokine receptor 3
Gene Aliases
CD182, CD183, CKR-L2, CMKAR3, GPR9, IP10-R, Mig-R, MigR
UniGene
Hs.198252
Gene Chromosome Location
Chr.X: 71615913-71618517 on Build GRCh38
Transcripts
NM_001142797.1 NM_001504.1 XM_005262256.3 XM_005262257.3 XM_017029435.1 XM_017029436.1

SNP Detail

SNP ID Chromosome Location
rs756780161 Chr.X: 71617178-71617178 on Build GRCh38
rs747049805 Chr.X: 71617255-71617255 on Build GRCh38
rs759889567 Chr.X: 71617151-71617151 on Build GRCh38
rs773243779 Chr.X: 71617254-71617254 on Build GRCh38
rs953556709 Chr.X: 71617271-71617271 on Build GRCh38
rs765668938 Chr.X: 71617173-71617173 on Build GRCh38
rs772183895 Chr.X: 71617098-71617098 on Build GRCh38
rs749047760 Chr.X: 71617241-71617241 on Build GRCh38
rs758505277 Chr.X: 71617229-71617229 on Build GRCh38
rs779802231 Chr.X: 71617230-71617230 on Build GRCh38
rs374595572 Chr.X: 71617244-71617244 on Build GRCh38
rs771157013 Chr.X: 71617258-71617258 on Build GRCh38
rs750143892 Chr.X: 71617208-71617208 on Build GRCh38
rs772188808 Chr.X: 71617247-71617247 on Build GRCh38
rs754230813 Chr.X: 71617135-71617135 on Build GRCh38
rs748102747 Chr.X: 71617245-71617245 on Build GRCh38
rs887473710 Chr.X: 71617136-71617136 on Build GRCh38
rs1015169579 Chr.X: 71617253-71617253 on Build GRCh38
rs760864060 Chr.X: 71617129-71617129 on Build GRCh38
rs753244136 Chr.X: 71617174-71617174 on Build GRCh38
rs866680405 Chr.X: 71617224-71617224 on Build GRCh38
rs139752678 Chr.X: 71617196-71617196 on Build GRCh38
rs773481155 Chr.X: 71617105-71617105 on Build GRCh38
rs372391641 Chr.X: 71617226-71617226 on Build GRCh38

View Full Product Details