Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 102838547-102838814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZIN1
Entrez Gene ID
51582
Gene Name
antizyme inhibitor 1
Gene Aliases
AZI, AZIA1, OAZI, OAZIN, ODC1L
UniGene
Hs.459106
Gene Chromosome Location
Chr.8: 102826302-102864200 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Gene Symbol
LOC107986873
Entrez Gene ID
107986873
Gene Name
forkhead box protein L2-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 102810302-102863067 on Build GRCh38
Transcripts
NM_001301668.1 NM_015878.5 NM_148174.3 XM_005250969.1 XM_011517126.1 XM_011517127.1 XM_011517128.2 XM_017013563.1 XM_017013564.1 XM_017013565.1 XM_017013566.1 XM_017014121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346026 c.431G>T p.R144L Chr.8: 102838762-102838762 on Build GRCh38
6609180 c.431G>A p.R144H Chr.8: 102838762-102838762 on Build GRCh38
6609182 c.416T>C p.L139S Chr.8: 102838777-102838777 on Build GRCh38
1094436 c.411T>G p.I137M Chr.8: 102838782-102838782 on Build GRCh38
1313555 c.417G>A p.L139L Chr.8: 102838776-102838776 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs62522602 Chr.8: 102838668-102838668 on Build GRCh38
rs960783354 Chr.8: 102838728-102838728 on Build GRCh38
rs750497005 Chr.8: 102838782-102838782 on Build GRCh38
rs772776067 Chr.8: 102838735-102838735 on Build GRCh38
rs567170313 Chr.8: 102838657-102838657 on Build GRCh38
rs1023647916 Chr.8: 102838765-102838765 on Build GRCh38
rs369562933 Chr.8: 102838709-102838709 on Build GRCh38
rs771693363 Chr.8: 102838725-102838725 on Build GRCh38
rs910909375 Chr.8: 102838647-102838647 on Build GRCh38
rs747740829 Chr.8: 102838716-102838716 on Build GRCh38
rs904051367 Chr.8: 102838703-102838703 on Build GRCh38
rs983844556 Chr.8: 102838577-102838577 on Build GRCh38
rs758885410 Chr.8: 102838658-102838658 on Build GRCh38
rs142294459 Chr.8: 102838763-102838763 on Build GRCh38
rs930458382 Chr.8: 102838619-102838619 on Build GRCh38
rs944421915 Chr.8: 102838652-102838652 on Build GRCh38
rs769248037 Chr.8: 102838729-102838729 on Build GRCh38
rs1052304560 Chr.8: 102838687-102838687 on Build GRCh38
rs543344671 Chr.8: 102838663-102838663 on Build GRCh38
rs942304562 Chr.8: 102838654-102838654 on Build GRCh38
rs184518812 Chr.8: 102838700-102838700 on Build GRCh38
rs750936028 Chr.8: 102838636-102838636 on Build GRCh38
rs759030712 Chr.8: 102838732-102838733 on Build GRCh38
rs377694303 Chr.8: 102838712-102838712 on Build GRCh38
rs759999261 Chr.8: 102838741-102838741 on Build GRCh38
rs765276991 Chr.8: 102838775-102838775 on Build GRCh38
rs935225757 Chr.8: 102838674-102838674 on Build GRCh38
rs764483089 Chr.8: 102838762-102838762 on Build GRCh38
rs763659703 Chr.8: 102838635-102838635 on Build GRCh38
rs768043887 Chr.8: 102838753-102838753 on Build GRCh38
rs781216249 Chr.8: 102838754-102838754 on Build GRCh38
rs761151867 Chr.8: 102838755-102838755 on Build GRCh38
rs952470347 Chr.8: 102838572-102838572 on Build GRCh38
rs373433614 Chr.8: 102838705-102838705 on Build GRCh38

View Full Product Details