Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168224202-168224463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5 XM_011532103.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1053014 c.4776G>C p.L1592L Chr.4: 168224291-168224291 on Build GRCh38
4904882 c.4748G>A p.R1583K Chr.4: 168224319-168224319 on Build GRCh38
4408093 c.4792G>A p.D1598N Chr.4: 168224275-168224275 on Build GRCh38
5011368 c.4795G>T p.D1599Y Chr.4: 168224272-168224272 on Build GRCh38
5584987 c.4782G>A p.G1594G Chr.4: 168224285-168224285 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960216493 Chr.4: 168224350-168224350 on Build GRCh38
rs771050410 Chr.4: 168224396-168224399 on Build GRCh38
rs369930154 Chr.4: 168224398-168224398 on Build GRCh38
rs372305019 Chr.4: 168224265-168224265 on Build GRCh38
rs181957271 Chr.4: 168224341-168224341 on Build GRCh38
rs2271712 Chr.4: 168224428-168224428 on Build GRCh38
rs957698915 Chr.4: 168224427-168224427 on Build GRCh38
rs759427776 Chr.4: 168224388-168224388 on Build GRCh38
rs755370439 Chr.4: 168224411-168224411 on Build GRCh38
rs766262357 Chr.4: 168224262-168224262 on Build GRCh38
rs888417416 Chr.4: 168224263-168224263 on Build GRCh38
rs1048875311 Chr.4: 168224260-168224260 on Build GRCh38
rs755442338 Chr.4: 168224342-168224342 on Build GRCh38
rs759515498 Chr.4: 168224275-168224275 on Build GRCh38
rs750654165 Chr.4: 168224319-168224319 on Build GRCh38
rs548035381 Chr.4: 168224276-168224276 on Build GRCh38
rs750553669 Chr.4: 168224401-168224401 on Build GRCh38
rs753812127 Chr.4: 168224284-168224284 on Build GRCh38
rs753231784 Chr.4: 168224433-168224433 on Build GRCh38
rs781727606 Chr.4: 168224348-168224348 on Build GRCh38
rs769268686 Chr.4: 168224243-168224243 on Build GRCh38
rs372165562 Chr.4: 168224406-168224406 on Build GRCh38
rs771846305 Chr.4: 168224391-168224391 on Build GRCh38
rs200137138 Chr.4: 168224335-168224335 on Build GRCh38
rs780376903 Chr.4: 168224334-168224334 on Build GRCh38
rs777203434 Chr.4: 168224367-168224367 on Build GRCh38
rs758681062 Chr.4: 168224331-168224331 on Build GRCh38
rs770747398 Chr.4: 168224379-168224379 on Build GRCh38
rs149475109 Chr.4: 168224250-168224250 on Build GRCh38
rs375521196 Chr.4: 168224351-168224351 on Build GRCh38
rs1024763401 Chr.4: 168224357-168224357 on Build GRCh38
rs757372390 Chr.4: 168224297-168224297 on Build GRCh38
rs2271711 Chr.4: 168224402-168224402 on Build GRCh38
rs774774632 Chr.4: 168224431-168224431 on Build GRCh38
rs748925536 Chr.4: 168224371-168224371 on Build GRCh38
rs765349264 Chr.4: 168224399-168224399 on Build GRCh38
rs143944090 Chr.4: 168224291-168224291 on Build GRCh38
rs766623463 Chr.4: 168224405-168224405 on Build GRCh38
rs569934179 Chr.4: 168224303-168224303 on Build GRCh38
rs112553793 Chr.4: 168224389-168224389 on Build GRCh38
rs769339815 Chr.4: 168224353-168224353 on Build GRCh38
rs200820384 Chr.4: 168224328-168224328 on Build GRCh38
rs529887389 Chr.4: 168224261-168224261 on Build GRCh38

View Full Product Details