Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220072801-220073026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPNT1
Entrez Gene ID
10380
Gene Name
3'(2'), 5'-bisphosphate nucleotidase 1
Gene Aliases
HEL20, PIP
UniGene
Hs.406134
Gene Chromosome Location
Chr.1: 220057482-220089916 on Build GRCh38
Transcripts
NM_001286149.1 NM_001286150.1 NM_001286151.1 NM_006085.5 XM_005272998.2 XM_005272999.4 XM_005273000.4 XM_005273004.2 XM_005273005.4 XM_006711114.3 XM_011509063.2 XM_011509064.2 XM_011509065.2 XM_017000043.1 XM_017000044.1 XM_017000045.1 XM_017000046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6061050 c.317C>G p.A106G Chr.1: 220072866-220072866 on Build GRCh38
1185738 c.247G>T p.D83Y Chr.1: 220072936-220072936 on Build GRCh38
6612403 c.306G>A p.S102S Chr.1: 220072877-220072877 on Build GRCh38
4665985 c.248A>C p.D83A Chr.1: 220072935-220072935 on Build GRCh38
4028375 c.246G>T p.V82V Chr.1: 220072937-220072937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918174759 Chr.1: 220072970-220072970 on Build GRCh38
rs777506639 Chr.1: 220072939-220072939 on Build GRCh38
rs749858545 Chr.1: 220072966-220072966 on Build GRCh38
rs999763134 Chr.1: 220072956-220072956 on Build GRCh38
rs757798490 Chr.1: 220072973-220072973 on Build GRCh38
rs758126497 Chr.1: 220072933-220072933 on Build GRCh38
rs760406349 Chr.1: 220072890-220072890 on Build GRCh38
rs190393666 Chr.1: 220072877-220072877 on Build GRCh38
rs750225034 Chr.1: 220072887-220072887 on Build GRCh38
rs776011878 Chr.1: 220072848-220072848 on Build GRCh38
rs766326710 Chr.1: 220072909-220072909 on Build GRCh38
rs746165724 Chr.1: 220072840-220072840 on Build GRCh38
rs762970937 Chr.1: 220072857-220072857 on Build GRCh38
rs540331407 Chr.1: 220072923-220072923 on Build GRCh38
rs200840033 Chr.1: 220072831-220072831 on Build GRCh38
rs780246975 Chr.1: 220072958-220072958 on Build GRCh38
rs753769635 Chr.1: 220072914-220072914 on Build GRCh38
rs770240390 Chr.1: 220072950-220072950 on Build GRCh38
rs746254440 Chr.1: 220072946-220072946 on Build GRCh38
rs574895553 Chr.1: 220072878-220072878 on Build GRCh38
rs754849098 Chr.1: 220072915-220072915 on Build GRCh38
rs992388161 Chr.1: 220072941-220072941 on Build GRCh38
rs374634264 Chr.1: 220072838-220072838 on Build GRCh38
rs199915166 Chr.1: 220072847-220072847 on Build GRCh38
rs984915370 Chr.1: 220072841-220072841 on Build GRCh38
rs768824567 Chr.1: 220072871-220072871 on Build GRCh38
rs769033797 Chr.1: 220072989-220072989 on Build GRCh38
rs752142105 Chr.1: 220072926-220072926 on Build GRCh38
rs372088611 Chr.1: 220072836-220072836 on Build GRCh38
rs749867028 Chr.1: 220072851-220072853 on Build GRCh38

View Full Product Details