Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137015718-137015985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM2
Entrez Gene ID
1129
Gene Name
cholinergic receptor muscarinic 2
Gene Aliases
HM2
UniGene
Hs.535891
Gene Chromosome Location
Chr.7: 136868652-137020255 on Build GRCh38
Transcripts
NM_000739.2 NM_001006626.1 NM_001006627.1 NM_001006628.1 NM_001006629.1 NM_001006630.1 NM_001006631.1 NM_001006632.1 XM_011515769.2 NR_046103.1

Gene Symbol
LOC349160
Entrez Gene ID
349160
Gene Name
uncharacterized LOC349160
Gene Aliases
-
UniGene
Hs.660713
Gene Chromosome Location
Chr.7: 136898773-137164341 on Build GRCh38
Transcripts
NM_000739.2 NM_001006626.1 NM_001006627.1 NM_001006628.1 NM_001006629.1 NM_001006630.1 NM_001006631.1 NM_001006632.1 XM_011515769.2 NR_046103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361235 c.1087C>A p.R363S Chr.7: 137015952-137015952 on Build GRCh38
1085936 c.1084G>A p.A362T Chr.7: 137015949-137015949 on Build GRCh38
5528671 c.898G>A p.E300K Chr.7: 137015763-137015763 on Build GRCh38
1448315 c.954G>A p.E318E Chr.7: 137015819-137015819 on Build GRCh38
5931990 c.910G>A p.D304N Chr.7: 137015775-137015775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778101866 Chr.7: 137015875-137015875 on Build GRCh38
rs779078351 Chr.7: 137015752-137015752 on Build GRCh38
rs760040127 Chr.7: 137015787-137015787 on Build GRCh38
rs1015524373 Chr.7: 137015906-137015906 on Build GRCh38
rs548996412 Chr.7: 137015957-137015957 on Build GRCh38
rs1034180842 Chr.7: 137015945-137015945 on Build GRCh38
rs950002962 Chr.7: 137015793-137015793 on Build GRCh38
rs774236034 Chr.7: 137015804-137015804 on Build GRCh38
rs753491070 Chr.7: 137015740-137015740 on Build GRCh38
rs267601307 Chr.7: 137015763-137015763 on Build GRCh38
rs758803826 Chr.7: 137015893-137015893 on Build GRCh38
rs143216855 Chr.7: 137015851-137015851 on Build GRCh38
rs374975955 Chr.7: 137015925-137015925 on Build GRCh38
rs60372903 Chr.7: 137015915-137015915 on Build GRCh38
rs770969121 Chr.7: 137015792-137015792 on Build GRCh38
rs748527931 Chr.7: 137015762-137015762 on Build GRCh38
rs773330734 Chr.7: 137015838-137015838 on Build GRCh38
rs201165506 Chr.7: 137015895-137015895 on Build GRCh38
rs200169935 Chr.7: 137015842-137015842 on Build GRCh38
rs915877356 Chr.7: 137015779-137015779 on Build GRCh38
rs892873306 Chr.7: 137015902-137015902 on Build GRCh38
rs764893658 Chr.7: 137015885-137015885 on Build GRCh38
rs149168625 Chr.7: 137015771-137015771 on Build GRCh38
rs866638024 Chr.7: 137015911-137015911 on Build GRCh38
rs767527482 Chr.7: 137015827-137015827 on Build GRCh38
rs769865494 Chr.7: 137015919-137015919 on Build GRCh38
rs755364385 Chr.7: 137015747-137015747 on Build GRCh38
rs747925877 Chr.7: 137015939-137015939 on Build GRCh38
rs200017653 Chr.7: 137015800-137015800 on Build GRCh38
rs772049510 Chr.7: 137015952-137015952 on Build GRCh38
rs139037603 Chr.7: 137015896-137015896 on Build GRCh38
rs755592247 Chr.7: 137015912-137015912 on Build GRCh38
rs147228075 Chr.7: 137015953-137015953 on Build GRCh38
rs781068894 Chr.7: 137015907-137015907 on Build GRCh38
rs77845403 Chr.7: 137015816-137015816 on Build GRCh38
rs955607223 Chr.7: 137015892-137015892 on Build GRCh38
rs761231662 Chr.7: 137015869-137015870 on Build GRCh38
rs747060290 Chr.7: 137015783-137015783 on Build GRCh38
rs772521126 Chr.7: 137015773-137015773 on Build GRCh38
rs778284565 Chr.7: 137015776-137015776 on Build GRCh38
rs146328962 Chr.7: 137015856-137015856 on Build GRCh38
rs776902210 Chr.7: 137015797-137015797 on Build GRCh38
rs143722099 Chr.7: 137015924-137015924 on Build GRCh38

View Full Product Details